Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50364545-50364810 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNA2D2
Entrez Gene ID
9254
Gene Name
calcium voltage-gated channel auxiliary subunit alpha2delta 2
Gene Aliases
CACNA2D
UniGene
Hs.476273
Gene Chromosome Location
Chr.3: 50362613-50504244 on Build GRCh38
Transcripts
NM_001005505.2 NM_001174051.2 NM_001291101.1 NM_006030.3 XM_005265581.4 XM_011534243.2 NR_111912.1

Gene Symbol
CYB561D2
Entrez Gene ID
11068
Gene Name
cytochrome b561 family member D2
Gene Aliases
101F6, TSP10, XXcos-LUCA11.4
UniGene
Hs.149443
Gene Chromosome Location
Chr.3: 50350695-50368197 on Build GRCh38
Transcripts
NM_001005505.2 NM_001174051.2 NM_001291101.1 NM_006030.3 XM_005265581.4 XM_011534243.2 NR_111912.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5776104 c.3419C>A p.S1140Y Chr.3: 50364679-50364679 on Build GRCh38
6241024 c.3387C>A p.P1129P Chr.3: 50364711-50364711 on Build GRCh38
1046515 c.3421C>T p.R1141C Chr.3: 50364677-50364677 on Build GRCh38
3595577 c.3327C>T p.F1109F Chr.3: 50364771-50364771 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs147278705 Chr.3: 50364728-50364728 on Build GRCh38
rs587614191 Chr.3: 50364619-50364619 on Build GRCh38
rs377531685 Chr.3: 50364777-50364777 on Build GRCh38
rs150832847 Chr.3: 50364700-50364700 on Build GRCh38
rs924946197 Chr.3: 50364753-50364753 on Build GRCh38
rs376300457 Chr.3: 50364674-50364674 on Build GRCh38
rs181614194 Chr.3: 50364752-50364752 on Build GRCh38
rs754681752 Chr.3: 50364715-50364715 on Build GRCh38
rs760852595 Chr.3: 50364622-50364622 on Build GRCh38
rs1008374141 Chr.3: 50364624-50364624 on Build GRCh38
rs374235053 Chr.3: 50364660-50364660 on Build GRCh38
rs765246193 Chr.3: 50364689-50364689 on Build GRCh38
rs1044915542 Chr.3: 50364634-50364634 on Build GRCh38
rs1020128289 Chr.3: 50364627-50364627 on Build GRCh38
rs772683542 Chr.3: 50364765-50364765 on Build GRCh38
rs917774309 Chr.3: 50364596-50364597 on Build GRCh38
rs903514227 Chr.3: 50364589-50364589 on Build GRCh38
rs772905217 Chr.3: 50364724-50364726 on Build GRCh38
rs1031978997 Chr.3: 50364741-50364741 on Build GRCh38
rs746798469 Chr.3: 50364710-50364710 on Build GRCh38
rs769485819 Chr.3: 50364738-50364738 on Build GRCh38
rs72934809 Chr.3: 50364749-50364749 on Build GRCh38
rs947781237 Chr.3: 50364571-50364571 on Build GRCh38
rs587740874 Chr.3: 50364616-50364616 on Build GRCh38
rs1014755697 Chr.3: 50364712-50364712 on Build GRCh38
rs145772306 Chr.3: 50364705-50364705 on Build GRCh38
rs1017724866 Chr.3: 50364725-50364725 on Build GRCh38
rs892639229 Chr.3: 50364618-50364618 on Build GRCh38
rs77176698 Chr.3: 50364729-50364729 on Build GRCh38
rs961872296 Chr.3: 50364726-50364726 on Build GRCh38
rs753961883 Chr.3: 50364684-50364684 on Build GRCh38
rs1044758811 Chr.3: 50364581-50364581 on Build GRCh38
rs941889396 Chr.3: 50364672-50364672 on Build GRCh38
rs988265792 Chr.3: 50364595-50364595 on Build GRCh38
rs987814006 Chr.3: 50364761-50364761 on Build GRCh38
rs186468159 Chr.3: 50364783-50364783 on Build GRCh38
rs890532196 Chr.3: 50364701-50364701 on Build GRCh38
rs936939902 Chr.3: 50364606-50364606 on Build GRCh38
rs150284749 Chr.3: 50364696-50364696 on Build GRCh38
rs761130363 Chr.3: 50364770-50364770 on Build GRCh38
rs947867877 Chr.3: 50364744-50364744 on Build GRCh38
rs369157817 Chr.3: 50364607-50364607 on Build GRCh38
rs777164486 Chr.3: 50364762-50364762 on Build GRCh38
rs984680991 Chr.3: 50364711-50364711 on Build GRCh38
rs757358420 Chr.3: 50364687-50364687 on Build GRCh38
rs775808130 Chr.3: 50364768-50364768 on Build GRCh38

View Full Product Details