Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 95866931-95867202 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC38
Entrez Gene ID
120935
Gene Name
coiled-coil domain containing 38
Gene Aliases
-
UniGene
Hs.210377
Gene Chromosome Location
Chr.12: 95867048-95943301 on Build GRCh38
Transcripts
NM_182496.2 XM_006719229.2 XM_011537883.2 XM_011537884.2 XM_011537886.2 XM_011537887.2 XM_011537888.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5376230 c.1586_1587GA>AT p.R529N Chr.12: 95867181-95867182 on Build GRCh38
317093 c.1583delG p.G528fs*20 Chr.12: 95867185-95867185 on Build GRCh38
944693 c.1682T>A p.F561Y Chr.12: 95867086-95867086 on Build GRCh38
5052006 c.1686delT p.F562fs*>2 Chr.12: 95867082-95867082 on Build GRCh38
2046415 c.1605A>C p.S535S Chr.12: 95867163-95867163 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs911783648 Chr.12: 95866960-95866960 on Build GRCh38
rs933595911 Chr.12: 95867156-95867156 on Build GRCh38
rs138854457 Chr.12: 95867145-95867145 on Build GRCh38
rs762709526 Chr.12: 95867139-95867139 on Build GRCh38
rs7960706 Chr.12: 95867169-95867169 on Build GRCh38
rs1053568056 Chr.12: 95867176-95867176 on Build GRCh38
rs142074919 Chr.12: 95867159-95867159 on Build GRCh38
rs1016376088 Chr.12: 95867066-95867066 on Build GRCh38
rs112454867 Chr.12: 95867143-95867143 on Build GRCh38
rs943083240 Chr.12: 95866967-95866967 on Build GRCh38
rs950835871 Chr.12: 95867011-95867011 on Build GRCh38
rs781382330 Chr.12: 95867062-95867062 on Build GRCh38
rs749881290 Chr.12: 95867041-95867041 on Build GRCh38
rs142643966 Chr.12: 95867116-95867116 on Build GRCh38
rs73227775 Chr.12: 95867108-95867108 on Build GRCh38
rs769501825 Chr.12: 95867082-95867082 on Build GRCh38
rs923480127 Chr.12: 95867155-95867155 on Build GRCh38
rs377664218 Chr.12: 95867058-95867058 on Build GRCh38
rs887302108 Chr.12: 95867138-95867138 on Build GRCh38
rs748431758 Chr.12: 95867079-95867079 on Build GRCh38
rs773024274 Chr.12: 95867107-95867110 on Build GRCh38
rs761417657 Chr.12: 95867036-95867036 on Build GRCh38
rs565250678 Chr.12: 95867102-95867102 on Build GRCh38
rs758914429 Chr.12: 95867042-95867042 on Build GRCh38
rs764752069 Chr.12: 95867037-95867037 on Build GRCh38
rs746895617 Chr.12: 95867158-95867158 on Build GRCh38
rs138606628 Chr.12: 95866997-95866997 on Build GRCh38
rs994511112 Chr.12: 95867089-95867089 on Build GRCh38
rs113446734 Chr.12: 95866977-95866977 on Build GRCh38
rs113263671 Chr.12: 95867111-95867111 on Build GRCh38
rs149303834 Chr.12: 95867090-95867090 on Build GRCh38
rs139730427 Chr.12: 95867170-95867170 on Build GRCh38
rs775552051 Chr.12: 95867153-95867153 on Build GRCh38
rs766938584 Chr.12: 95867043-95867043 on Build GRCh38
rs751997057 Chr.12: 95867051-95867051 on Build GRCh38

View Full Product Details