Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 159884363-159884636 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP45
Entrez Gene ID
25790
Gene Name
cilia and flagella associated protein 45
Gene Aliases
CCDC19, NESG1
UniGene
Hs.647705
Gene Chromosome Location
Chr.1: 159872364-159900116 on Build GRCh38
Transcripts
NM_012337.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5844142 c.836G>A p.R279Q Chr.1: 159884497-159884497 on Build GRCh38
3476632 c.844G>A p.E282K Chr.1: 159884489-159884489 on Build GRCh38
5844143 c.787delG p.E263fs*36 Chr.1: 159884546-159884546 on Build GRCh38
5889464 c.768-4C>T p.? Chr.1: 159884569-159884569 on Build GRCh38
336604 c.819G>A p.S273S Chr.1: 159884514-159884514 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749483000 Chr.1: 159884409-159884409 on Build GRCh38
rs752665362 Chr.1: 159884396-159884396 on Build GRCh38
rs757380651 Chr.1: 159884589-159884589 on Build GRCh38
rs767864203 Chr.1: 159884497-159884497 on Build GRCh38
rs778565474 Chr.1: 159884413-159884413 on Build GRCh38
rs547705787 Chr.1: 159884487-159884487 on Build GRCh38
rs753278916 Chr.1: 159884498-159884498 on Build GRCh38
rs765009883 Chr.1: 159884483-159884483 on Build GRCh38
rs972781859 Chr.1: 159884529-159884529 on Build GRCh38
rs775884442 Chr.1: 159884581-159884581 on Build GRCh38
rs151219155 Chr.1: 159884515-159884515 on Build GRCh38
rs764137419 Chr.1: 159884514-159884514 on Build GRCh38
rs115172513 Chr.1: 159884572-159884572 on Build GRCh38
rs753806120 Chr.1: 159884487-159884489 on Build GRCh38
rs767605282 Chr.1: 159884558-159884558 on Build GRCh38
rs1054606983 Chr.1: 159884595-159884595 on Build GRCh38
rs745600197 Chr.1: 159884525-159884525 on Build GRCh38
rs761164701 Chr.1: 159884583-159884583 on Build GRCh38
rs779208743 Chr.1: 159884519-159884519 on Build GRCh38
rs41264829 Chr.1: 159884518-159884518 on Build GRCh38
rs183286654 Chr.1: 159884385-159884385 on Build GRCh38
rs753856046 Chr.1: 159884587-159884587 on Build GRCh38
rs16842789 Chr.1: 159884461-159884461 on Build GRCh38
rs756209120 Chr.1: 159884397-159884397 on Build GRCh38
rs758594790 Chr.1: 159884598-159884598 on Build GRCh38
rs374764461 Chr.1: 159884586-159884586 on Build GRCh38
rs772944551 Chr.1: 159884564-159884564 on Build GRCh38
rs768247708 Chr.1: 159884463-159884463 on Build GRCh38
rs765505059 Chr.1: 159884394-159884394 on Build GRCh38
rs938328112 Chr.1: 159884544-159884544 on Build GRCh38
rs767892647 Chr.1: 159884574-159884574 on Build GRCh38
rs779750153 Chr.1: 159884538-159884538 on Build GRCh38
rs186196147 Chr.1: 159884597-159884597 on Build GRCh38
rs990295458 Chr.1: 159884474-159884474 on Build GRCh38
rs376776397 Chr.1: 159884469-159884469 on Build GRCh38
rs980672681 Chr.1: 159884578-159884578 on Build GRCh38
rs566322359 Chr.1: 159884547-159884547 on Build GRCh38
rs370229782 Chr.1: 159884557-159884557 on Build GRCh38
rs1044596574 Chr.1: 159884422-159884422 on Build GRCh38
rs777934453 Chr.1: 159884402-159884402 on Build GRCh38
rs745680245 Chr.1: 159884416-159884416 on Build GRCh38
rs2501325 Chr.1: 159884602-159884602 on Build GRCh38
rs759961532 Chr.1: 159884493-159884493 on Build GRCh38
rs771846034 Chr.1: 159884418-159884418 on Build GRCh38
rs375491975 Chr.1: 159884511-159884511 on Build GRCh38
rs746918360 Chr.1: 159884539-159884539 on Build GRCh38
rs563332239 Chr.1: 159884384-159884384 on Build GRCh38
rs1054637350 Chr.1: 159884441-159884441 on Build GRCh38
rs961340993 Chr.1: 159884521-159884521 on Build GRCh38
rs200874439 Chr.1: 159884484-159884484 on Build GRCh38
rs764264921 Chr.1: 159884466-159884466 on Build GRCh38
rs757531174 Chr.1: 159884410-159884410 on Build GRCh38
rs374481152 Chr.1: 159884431-159884431 on Build GRCh38
rs372564105 Chr.1: 159884476-159884476 on Build GRCh38
rs758241535 Chr.1: 159884532-159884532 on Build GRCh38

View Full Product Details