Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 58323356-58323619 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARID4A
Entrez Gene ID
5926
Gene Name
AT-rich interaction domain 4A
Gene Aliases
RBBP-1, RBBP1, RBP-1, RBP1
UniGene
Hs.161000
Gene Chromosome Location
Chr.14: 58298385-58373733 on Build GRCh38
Transcripts
NM_002892.3 NM_023000.2 NM_023001.2 XM_005267964.2 XM_005267965.2 XM_017021560.1 XM_017021561.1 XM_017021562.1 XM_017021563.1 XM_017021564.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5654282 c.457G>C p.D153H Chr.14: 58323492-58323492 on Build GRCh38
4051440 c.502C>T p.R168C Chr.14: 58323537-58323537 on Build GRCh38
3361172 c.517G>T p.E173* Chr.14: 58323552-58323552 on Build GRCh38
698271 c.501G>T p.K167N Chr.14: 58323536-58323536 on Build GRCh38
4663283 c.535_536insTT p.S180fs*1 Chr.14: 58323570-58323571 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755821108 Chr.14: 58323585-58323585 on Build GRCh38
rs781095768 Chr.14: 58323502-58323502 on Build GRCh38
rs368715644 Chr.14: 58323541-58323541 on Build GRCh38
rs189434559 Chr.14: 58323463-58323463 on Build GRCh38
rs892212539 Chr.14: 58323527-58323527 on Build GRCh38
rs762414235 Chr.14: 58323434-58323434 on Build GRCh38
rs781236840 Chr.14: 58323581-58323581 on Build GRCh38
rs570701333 Chr.14: 58323464-58323464 on Build GRCh38
rs780434744 Chr.14: 58323485-58323485 on Build GRCh38
rs1053500733 Chr.14: 58323525-58323525 on Build GRCh38
rs951326951 Chr.14: 58323443-58323446 on Build GRCh38
rs773098388 Chr.14: 58323533-58323533 on Build GRCh38
rs765867768 Chr.14: 58323555-58323555 on Build GRCh38
rs755274345 Chr.14: 58323580-58323580 on Build GRCh38
rs200071799 Chr.14: 58323519-58323519 on Build GRCh38
rs139306325 Chr.14: 58323589-58323589 on Build GRCh38
rs936432419 Chr.14: 58323476-58323476 on Build GRCh38
rs774540026 Chr.14: 58323423-58323423 on Build GRCh38
rs747159680 Chr.14: 58323487-58323487 on Build GRCh38
rs747869988 Chr.14: 58323504-58323504 on Build GRCh38
rs751166987 Chr.14: 58323453-58323453 on Build GRCh38
rs767920033 Chr.14: 58323570-58323570 on Build GRCh38
rs757841211 Chr.14: 58323465-58323465 on Build GRCh38
rs751673938 Chr.14: 58323578-58323578 on Build GRCh38
rs748822451 Chr.14: 58323534-58323534 on Build GRCh38
rs760962942 Chr.14: 58323457-58323457 on Build GRCh38
rs768753599 Chr.14: 58323494-58323494 on Build GRCh38
rs748002398 Chr.14: 58323584-58323584 on Build GRCh38
rs770373766 Chr.14: 58323537-58323537 on Build GRCh38
rs758555881 Chr.14: 58323566-58323566 on Build GRCh38
rs750306635 Chr.14: 58323501-58323503 on Build GRCh38
rs762052890 Chr.14: 58323546-58323546 on Build GRCh38
rs906463672 Chr.14: 58323588-58323588 on Build GRCh38
rs371437846 Chr.14: 58323474-58323474 on Build GRCh38
rs146397124 Chr.14: 58323540-58323540 on Build GRCh38
rs766631347 Chr.14: 58323573-58323573 on Build GRCh38
rs766056261 Chr.14: 58323450-58323450 on Build GRCh38
rs943587400 Chr.14: 58323427-58323427 on Build GRCh38
rs773999866 Chr.14: 58323538-58323538 on Build GRCh38
rs749158890 Chr.14: 58323590-58323590 on Build GRCh38
rs758422159 Chr.14: 58323546-58323548 on Build GRCh38
rs758502852 Chr.14: 58323478-58323478 on Build GRCh38
rs750902132 Chr.14: 58323558-58323558 on Build GRCh38
rs140731982 Chr.14: 58323467-58323467 on Build GRCh38
rs374197803 Chr.14: 58323524-58323524 on Build GRCh38

View Full Product Details