Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 44052573-44052846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACCSL
Entrez Gene ID
390110
Gene Name
1-aminocyclopropane-1-carboxylate synthase homolog (inactive) like
Gene Aliases
-
UniGene
Hs.558851
Gene Chromosome Location
Chr.11: 43920987-44060667 on Build GRCh38
Transcripts
NM_001031854.2 XM_017017714.1 XM_017017715.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3676158 c.795C>T p.P265P Chr.11: 44052684-44052684 on Build GRCh38
4768925 c.841G>C p.E281Q Chr.11: 44052730-44052730 on Build GRCh38
7414831 c.791C>A p.A264D Chr.11: 44052680-44052680 on Build GRCh38
5838525 c.783delG p.V262fs*21 Chr.11: 44052672-44052672 on Build GRCh38
4510882 c.850C>T p.P284S Chr.11: 44052739-44052739 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377326692 Chr.11: 44052712-44052712 on Build GRCh38
rs756675485 Chr.11: 44052599-44052599 on Build GRCh38
rs765330940 Chr.11: 44052673-44052673 on Build GRCh38
rs759364468 Chr.11: 44052738-44052738 on Build GRCh38
rs772402298 Chr.11: 44052605-44052605 on Build GRCh38
rs767494956 Chr.11: 44052649-44052649 on Build GRCh38
rs747890227 Chr.11: 44052689-44052689 on Build GRCh38
rs769225471 Chr.11: 44052707-44052707 on Build GRCh38
rs772787198 Chr.11: 44052617-44052617 on Build GRCh38
rs771970035 Chr.11: 44052745-44052745 on Build GRCh38
rs200113453 Chr.11: 44052672-44052672 on Build GRCh38
rs753995174 Chr.11: 44052652-44052652 on Build GRCh38
rs772960780 Chr.11: 44052822-44052822 on Build GRCh38
rs753468171 Chr.11: 44052655-44052655 on Build GRCh38
rs1038154309 Chr.11: 44052650-44052650 on Build GRCh38
rs767834149 Chr.11: 44052790-44052790 on Build GRCh38
rs77666758 Chr.11: 44052753-44052753 on Build GRCh38
rs544849804 Chr.11: 44052702-44052702 on Build GRCh38
rs764800733 Chr.11: 44052719-44052720 on Build GRCh38
rs763092912 Chr.11: 44052774-44052774 on Build GRCh38
rs754644938 Chr.11: 44052696-44052696 on Build GRCh38
rs936116610 Chr.11: 44052759-44052759 on Build GRCh38
rs770697296 Chr.11: 44052635-44052635 on Build GRCh38
rs766203957 Chr.11: 44052627-44052628 on Build GRCh38
rs766587982 Chr.11: 44052775-44052775 on Build GRCh38
rs760687487 Chr.11: 44052756-44052756 on Build GRCh38
rs780298251 Chr.11: 44052683-44052683 on Build GRCh38
rs774019979 Chr.11: 44052726-44052726 on Build GRCh38
rs755381266 Chr.11: 44052789-44052789 on Build GRCh38
rs193251654 Chr.11: 44052762-44052762 on Build GRCh38
rs752084816 Chr.11: 44052727-44052729 on Build GRCh38
rs185712469 Chr.11: 44052782-44052782 on Build GRCh38
rs748949652 Chr.11: 44052713-44052713 on Build GRCh38
rs189288309 Chr.11: 44052642-44052642 on Build GRCh38
rs773288959 Chr.11: 44052772-44052772 on Build GRCh38
rs35617980 Chr.11: 44052659-44052660 on Build GRCh38
rs770558690 Chr.11: 44052717-44052717 on Build GRCh38
rs781625008 Chr.11: 44052701-44052701 on Build GRCh38
rs750679229 Chr.11: 44052680-44052680 on Build GRCh38
rs755513760 Chr.11: 44052694-44052694 on Build GRCh38
rs753267297 Chr.11: 44052804-44052804 on Build GRCh38
rs758635417 Chr.11: 44052682-44052682 on Build GRCh38
rs747614258 Chr.11: 44052616-44052616 on Build GRCh38
rs1002942599 Chr.11: 44052817-44052817 on Build GRCh38
rs751858410 Chr.11: 44052692-44052692 on Build GRCh38
rs1018416635 Chr.11: 44052665-44052665 on Build GRCh38

View Full Product Details