Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 13419445-13419692 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD10
Entrez Gene ID
84280
Gene Name
BTB domain containing 10
Gene Aliases
GMRP-1, GMRP1
UniGene
Hs.332382
Gene Chromosome Location
Chr.11: 13387996-13463297 on Build GRCh38
Transcripts
NM_001297741.1 NM_001297742.1 NM_032320.6 XM_017018405.1 XM_017018406.1 XM_017018407.1 XM_017018408.1 XM_017018409.1 XM_017018410.1 XM_017018411.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3368308 c.557C>T p.T186I Chr.11: 13419487-13419487 on Build GRCh38
338431 c.454G>A p.V152I Chr.11: 13419590-13419590 on Build GRCh38
4019586 c.414T>G p.S138R Chr.11: 13419630-13419630 on Build GRCh38
6274223 c.566C>A p.P189Q Chr.11: 13419478-13419478 on Build GRCh38
3397581 c.471A>G p.K157K Chr.11: 13419573-13419573 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754954191 Chr.11: 13419643-13419643 on Build GRCh38
rs768183636 Chr.11: 13419534-13419534 on Build GRCh38
rs533959399 Chr.11: 13419573-13419573 on Build GRCh38
rs760109036 Chr.11: 13419665-13419665 on Build GRCh38
rs771169663 Chr.11: 13419661-13419661 on Build GRCh38
rs774906483 Chr.11: 13419664-13419664 on Build GRCh38
rs776156456 Chr.11: 13419542-13419542 on Build GRCh38
rs779864912 Chr.11: 13419510-13419510 on Build GRCh38
rs746915279 Chr.11: 13419641-13419641 on Build GRCh38
rs200018848 Chr.11: 13419556-13419556 on Build GRCh38
rs761431759 Chr.11: 13419543-13419543 on Build GRCh38
rs779997318 Chr.11: 13419617-13419617 on Build GRCh38
rs764230713 Chr.11: 13419605-13419605 on Build GRCh38
rs745815391 Chr.11: 13419552-13419552 on Build GRCh38
rs756403765 Chr.11: 13419590-13419590 on Build GRCh38
rs979546409 Chr.11: 13419550-13419550 on Build GRCh38
rs771981722 Chr.11: 13419503-13419503 on Build GRCh38
rs772752682 Chr.11: 13419656-13419656 on Build GRCh38
rs769421195 Chr.11: 13419653-13419653 on Build GRCh38
rs752885456 Chr.11: 13419589-13419589 on Build GRCh38
rs749625240 Chr.11: 13419658-13419658 on Build GRCh38
rs370433830 Chr.11: 13419531-13419531 on Build GRCh38
rs373440669 Chr.11: 13419649-13419649 on Build GRCh38
rs745720513 Chr.11: 13419494-13419494 on Build GRCh38
rs199658475 Chr.11: 13419549-13419549 on Build GRCh38
rs763231990 Chr.11: 13419559-13419559 on Build GRCh38
rs373290906 Chr.11: 13419470-13419470 on Build GRCh38
rs746469885 Chr.11: 13419519-13419519 on Build GRCh38
rs766861687 Chr.11: 13419563-13419563 on Build GRCh38
rs767685098 Chr.11: 13419586-13419586 on Build GRCh38
rs377504825 Chr.11: 13419501-13419501 on Build GRCh38
rs747722217 Chr.11: 13419652-13419652 on Build GRCh38
rs7114113 Chr.11: 13419579-13419579 on Build GRCh38
rs34185489 Chr.11: 13419611-13419611 on Build GRCh38
rs750179233 Chr.11: 13419608-13419608 on Build GRCh38

View Full Product Details