Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 148613977-148614248 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf33
Entrez Gene ID
202865
Gene Name
chromosome 7 open reading frame 33
Gene Aliases
-
UniGene
Hs.352212
Gene Chromosome Location
Chr.7: 148590565-148615860 on Build GRCh38
Transcripts
NM_145304.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3674858 c.284C>A p.P95Q Chr.7: 148614121-148614121 on Build GRCh38
3635727 c.357C>T p.G119G Chr.7: 148614194-148614194 on Build GRCh38
6176711 c.207G>C p.K69N Chr.7: 148614044-148614044 on Build GRCh38
6109198 c.234G>T p.R78R Chr.7: 148614071-148614071 on Build GRCh38
7244541 c.307C>G p.P103A Chr.7: 148614144-148614144 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201586129 Chr.7: 148614216-148614216 on Build GRCh38
rs151073020 Chr.7: 148614121-148614121 on Build GRCh38
rs752097738 Chr.7: 148614166-148614166 on Build GRCh38
rs73463436 Chr.7: 148614148-148614148 on Build GRCh38
rs748063011 Chr.7: 148614017-148614017 on Build GRCh38
rs759718311 Chr.7: 148614056-148614056 on Build GRCh38
rs566641914 Chr.7: 148614069-148614069 on Build GRCh38
rs370470523 Chr.7: 148614160-148614160 on Build GRCh38
rs776790488 Chr.7: 148614040-148614040 on Build GRCh38
rs1000619918 Chr.7: 148614149-148614149 on Build GRCh38
rs778780697 Chr.7: 148614138-148614138 on Build GRCh38
rs763194440 Chr.7: 148614171-148614171 on Build GRCh38
rs201257705 Chr.7: 148614027-148614027 on Build GRCh38
rs759588680 Chr.7: 148614070-148614070 on Build GRCh38
rs766453541 Chr.7: 148614026-148614027 on Build GRCh38
rs192458848 Chr.7: 148614109-148614109 on Build GRCh38
rs546618103 Chr.7: 148614063-148614063 on Build GRCh38
rs992784713 Chr.7: 148614167-148614167 on Build GRCh38
rs6965813 Chr.7: 148614122-148614122 on Build GRCh38
rs754614935 Chr.7: 148614221-148614221 on Build GRCh38
rs534493236 Chr.7: 148614220-148614220 on Build GRCh38
rs529999126 Chr.7: 148614019-148614019 on Build GRCh38
rs201949537 Chr.7: 148614062-148614062 on Build GRCh38
rs777921352 Chr.7: 148614150-148614150 on Build GRCh38
rs768071614 Chr.7: 148614107-148614107 on Build GRCh38
rs370035033 Chr.7: 148614010-148614010 on Build GRCh38
rs751926009 Chr.7: 148614098-148614098 on Build GRCh38
rs996413461 Chr.7: 148614103-148614103 on Build GRCh38
rs762128590 Chr.7: 148614016-148614019 on Build GRCh38
rs779553473 Chr.7: 148614115-148614115 on Build GRCh38
rs768879414 Chr.7: 148614186-148614186 on Build GRCh38
rs11434056 Chr.7: 148614023-148614024 on Build GRCh38
rs762234030 Chr.7: 148614204-148614204 on Build GRCh38
rs201426641 Chr.7: 148614207-148614207 on Build GRCh38
rs199640449 Chr.7: 148614215-148614215 on Build GRCh38
rs753987094 Chr.7: 148614031-148614032 on Build GRCh38
rs373786504 Chr.7: 148614099-148614099 on Build GRCh38
rs200989751 Chr.7: 148614070-148614070 on Build GRCh38
rs375826386 Chr.7: 148614152-148614152 on Build GRCh38
rs760894231 Chr.7: 148614025-148614027 on Build GRCh38
rs184505375 Chr.7: 148614203-148614203 on Build GRCh38
rs772445613 Chr.7: 148614015-148614016 on Build GRCh38
rs201498451 Chr.7: 148614048-148614048 on Build GRCh38
rs755693750 Chr.7: 148614113-148614113 on Build GRCh38
rs775454297 Chr.7: 148614162-148614162 on Build GRCh38
rs773922460 Chr.7: 148614075-148614075 on Build GRCh38

View Full Product Details