Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33573731-33573970 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAK1
Entrez Gene ID
578
Gene Name
BCL2 antagonist/killer 1
Gene Aliases
BAK, BAK-LIKE, BCL2L7, CDN1
UniGene
Hs.485139
Gene Chromosome Location
Chr.6: 33572546-33580293 on Build GRCh38
Transcripts
NM_001188.3 XM_011514779.2 XM_011514780.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
451315 c.617G>A p.R206Q Chr.6: 33573822-33573822 on Build GRCh38
231828 c.602G>A p.G201D Chr.6: 33573837-33573837 on Build GRCh38
1443831 c.624C>A p.F208L Chr.6: 33573815-33573815 on Build GRCh38
6034735 c.577G>A p.V193M Chr.6: 33573862-33573862 on Build GRCh38
7075774 c.574C>G p.L192V Chr.6: 33573865-33573865 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757957290 Chr.6: 33573947-33573947 on Build GRCh38
rs767030876 Chr.6: 33573777-33573777 on Build GRCh38
rs753158141 Chr.6: 33573944-33573944 on Build GRCh38
rs777128029 Chr.6: 33573825-33573825 on Build GRCh38
rs754404299 Chr.6: 33573927-33573927 on Build GRCh38
rs778415235 Chr.6: 33573924-33573926 on Build GRCh38
rs755412880 Chr.6: 33573792-33573792 on Build GRCh38
rs778565612 Chr.6: 33573795-33573795 on Build GRCh38
rs766939211 Chr.6: 33573925-33573925 on Build GRCh38
rs368163314 Chr.6: 33573938-33573938 on Build GRCh38
rs754210197 Chr.6: 33573787-33573787 on Build GRCh38
rs372889857 Chr.6: 33573868-33573868 on Build GRCh38
rs769479388 Chr.6: 33573869-33573869 on Build GRCh38
rs761926026 Chr.6: 33573901-33573901 on Build GRCh38
rs772385015 Chr.6: 33573907-33573907 on Build GRCh38
rs200704036 Chr.6: 33573839-33573839 on Build GRCh38
rs513349 Chr.6: 33573942-33573942 on Build GRCh38
rs376265958 Chr.6: 33573796-33573796 on Build GRCh38
rs781130899 Chr.6: 33573861-33573861 on Build GRCh38
rs370944014 Chr.6: 33573823-33573823 on Build GRCh38
rs181393571 Chr.6: 33573756-33573756 on Build GRCh38
rs7341238 Chr.6: 33573776-33573776 on Build GRCh38
rs773391910 Chr.6: 33573912-33573912 on Build GRCh38
rs186474429 Chr.6: 33573892-33573892 on Build GRCh38
rs746563887 Chr.6: 33573846-33573846 on Build GRCh38
rs760867106 Chr.6: 33573913-33573913 on Build GRCh38
rs745605734 Chr.6: 33573865-33573865 on Build GRCh38
rs775422894 Chr.6: 33573870-33573870 on Build GRCh38
rs372327476 Chr.6: 33573838-33573838 on Build GRCh38
rs770817553 Chr.6: 33573860-33573860 on Build GRCh38

View Full Product Details