Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 76985548-76985819 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACER3
Entrez Gene ID
55331
Gene Name
alkaline ceramidase 3
Gene Aliases
APHC, PHCA
UniGene
Hs.23862
Gene Chromosome Location
Chr.11: 76860873-77026797 on Build GRCh38
Transcripts
NM_001300953.1 NM_001300954.1 NM_001300955.1 NM_018367.6 XM_011545151.2 XM_011545152.2 XM_011545153.2 XM_017017987.1 XM_017017988.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5799553 c.339C>G p.I113M Chr.11: 76985661-76985661 on Build GRCh38
429883 c.336G>T p.K112N Chr.11: 76985658-76985658 on Build GRCh38
6421283 c.341A>G p.K114R Chr.11: 76985663-76985663 on Build GRCh38
196434 c.390A>G p.L130L Chr.11: 76985712-76985712 on Build GRCh38
4658812 c.365delT p.F123fs*3 Chr.11: 76985687-76985687 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs906859466 Chr.11: 76985742-76985742 on Build GRCh38
rs373296234 Chr.11: 76985614-76985614 on Build GRCh38
rs747484955 Chr.11: 76985583-76985583 on Build GRCh38
rs898100069 Chr.11: 76985647-76985647 on Build GRCh38
rs374372817 Chr.11: 76985746-76985746 on Build GRCh38
rs200603070 Chr.11: 76985733-76985733 on Build GRCh38
rs145982324 Chr.11: 76985724-76985724 on Build GRCh38
rs767901136 Chr.11: 76985732-76985732 on Build GRCh38
rs185775967 Chr.11: 76985770-76985770 on Build GRCh38
rs17135310 Chr.11: 76985590-76985590 on Build GRCh38
rs777832951 Chr.11: 76985649-76985649 on Build GRCh38
rs111412584 Chr.11: 76985606-76985606 on Build GRCh38
rs755231048 Chr.11: 76985757-76985757 on Build GRCh38
rs376143519 Chr.11: 76985639-76985639 on Build GRCh38
rs776671988 Chr.11: 76985632-76985632 on Build GRCh38
rs201991223 Chr.11: 76985609-76985610 on Build GRCh38
rs867663483 Chr.11: 76985598-76985598 on Build GRCh38
rs768711159 Chr.11: 76985626-76985629 on Build GRCh38
rs746486966 Chr.11: 76985663-76985663 on Build GRCh38
rs376328936 Chr.11: 76985619-76985619 on Build GRCh38
rs1037833312 Chr.11: 76985620-76985623 on Build GRCh38
rs369879809 Chr.11: 76985610-76985610 on Build GRCh38
rs752426317 Chr.11: 76985774-76985774 on Build GRCh38
rs753997229 Chr.11: 76985755-76985755 on Build GRCh38
rs778803515 Chr.11: 76985615-76985615 on Build GRCh38
rs759145370 Chr.11: 76985703-76985703 on Build GRCh38
rs79316237 Chr.11: 76985680-76985680 on Build GRCh38
rs769455875 Chr.11: 76985712-76985712 on Build GRCh38
rs781514022 Chr.11: 76985634-76985634 on Build GRCh38
rs774669854 Chr.11: 76985715-76985715 on Build GRCh38
rs761091992 Chr.11: 76985747-76985747 on Build GRCh38
rs370822453 Chr.11: 76985694-76985694 on Build GRCh38

View Full Product Details