Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 757295-757556 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MISP
Entrez Gene ID
126353
Gene Name
mitotic spindle positioning
Gene Aliases
C19orf21, MISP1
UniGene
Hs.439180
Gene Chromosome Location
Chr.19: 748413-764319 on Build GRCh38
Transcripts
NM_173481.3 NR_135168.1 XM_011527685.2 XM_011527686.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6282568 c.597G>T p.Q199H Chr.19: 757543-757543 on Build GRCh38
7252693 c.513G>A p.R171R Chr.19: 757459-757459 on Build GRCh38
5589897 c.547G>A p.E183K Chr.19: 757493-757493 on Build GRCh38
7377190 c.436G>T p.V146F Chr.19: 757382-757382 on Build GRCh38
4439337 c.483C>T p.L161L Chr.19: 757429-757429 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372409482 Chr.19: 757482-757482 on Build GRCh38
rs779862605 Chr.19: 757429-757429 on Build GRCh38
rs745862363 Chr.19: 757380-757380 on Build GRCh38
rs776972496 Chr.19: 757400-757400 on Build GRCh38
rs367991450 Chr.19: 757367-757367 on Build GRCh38
rs374036064 Chr.19: 757425-757425 on Build GRCh38
rs774073962 Chr.19: 757323-757323 on Build GRCh38
rs757352679 Chr.19: 757373-757373 on Build GRCh38
rs756144674 Chr.19: 757427-757427 on Build GRCh38
rs763267363 Chr.19: 757469-757469 on Build GRCh38
rs147697749 Chr.19: 757337-757337 on Build GRCh38
rs778897192 Chr.19: 757487-757487 on Build GRCh38
rs35484463 Chr.19: 757483-757483 on Build GRCh38
rs867869366 Chr.19: 757434-757434 on Build GRCh38
rs751813606 Chr.19: 757417-757417 on Build GRCh38
rs754786153 Chr.19: 757484-757484 on Build GRCh38
rs767724228 Chr.19: 757523-757523 on Build GRCh38
rs35090443 Chr.19: 757354-757354 on Build GRCh38
rs1000677074 Chr.19: 757463-757463 on Build GRCh38
rs375696497 Chr.19: 757492-757492 on Build GRCh38
rs746467418 Chr.19: 757501-757501 on Build GRCh38
rs988439292 Chr.19: 757363-757363 on Build GRCh38
rs937754023 Chr.19: 757386-757386 on Build GRCh38
rs768359832 Chr.19: 757391-757391 on Build GRCh38
rs368732999 Chr.19: 757533-757533 on Build GRCh38
rs368789822 Chr.19: 757480-757480 on Build GRCh38
rs573800828 Chr.19: 757382-757382 on Build GRCh38
rs201623422 Chr.19: 757319-757319 on Build GRCh38
rs748946118 Chr.19: 757387-757387 on Build GRCh38
rs543367056 Chr.19: 757472-757472 on Build GRCh38
rs78823735 Chr.19: 757465-757465 on Build GRCh38
rs867122964 Chr.19: 757520-757520 on Build GRCh38
rs372950958 Chr.19: 757447-757447 on Build GRCh38
rs757256662 Chr.19: 757418-757418 on Build GRCh38
rs780954431 Chr.19: 757374-757374 on Build GRCh38
rs3746172 Chr.19: 757399-757399 on Build GRCh38
rs546859511 Chr.19: 757499-757499 on Build GRCh38
rs145204788 Chr.19: 757461-757461 on Build GRCh38
rs777823655 Chr.19: 757366-757366 on Build GRCh38
rs376883941 Chr.19: 757530-757530 on Build GRCh38
rs3746173 Chr.19: 757412-757412 on Build GRCh38
rs375452577 Chr.19: 757468-757468 on Build GRCh38
rs750314792 Chr.19: 757426-757426 on Build GRCh38
rs371384413 Chr.19: 757466-757466 on Build GRCh38
rs762157919 Chr.19: 757473-757473 on Build GRCh38
rs920970624 Chr.19: 757405-757405 on Build GRCh38
rs78455638 Chr.19: 757509-757509 on Build GRCh38
rs369591220 Chr.19: 757515-757515 on Build GRCh38
rs141357485 Chr.19: 757534-757534 on Build GRCh38
rs35869813 Chr.19: 757321-757321 on Build GRCh38
rs765503068 Chr.19: 757410-757410 on Build GRCh38
rs774158068 Chr.19: 757392-757392 on Build GRCh38
rs749031887 Chr.19: 757439-757439 on Build GRCh38
rs537290556 Chr.19: 757365-757365 on Build GRCh38
rs371906123 Chr.19: 757516-757516 on Build GRCh38
rs962776128 Chr.19: 757508-757508 on Build GRCh38
rs764486603 Chr.19: 757470-757470 on Build GRCh38
rs370299354 Chr.19: 757370-757370 on Build GRCh38
rs1033901456 Chr.19: 757325-757325 on Build GRCh38
rs151258719 Chr.19: 757411-757411 on Build GRCh38
rs760134873 Chr.19: 757408-757408 on Build GRCh38
rs758135556 Chr.19: 757498-757498 on Build GRCh38
rs770380725 Chr.19: 757502-757502 on Build GRCh38
rs868722836 Chr.19: 757457-757457 on Build GRCh38
rs775161053 Chr.19: 757387-757387 on Build GRCh38
rs151208927 Chr.19: 757320-757320 on Build GRCh38
rs747842095 Chr.19: 757455-757455 on Build GRCh38
rs769454588 Chr.19: 757381-757381 on Build GRCh38
rs367585510 Chr.19: 757444-757444 on Build GRCh38
rs536955289 Chr.19: 757334-757334 on Build GRCh38
rs759779671 Chr.19: 757338-757338 on Build GRCh38
rs866613262 Chr.19: 757403-757403 on Build GRCh38
rs767493982 Chr.19: 757475-757475 on Build GRCh38
rs377309090 Chr.19: 757476-757476 on Build GRCh38
rs765670492 Chr.19: 757343-757343 on Build GRCh38

View Full Product Details