Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 97583475-97583702 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD2
Entrez Gene ID
26287
Gene Name
ankyrin repeat domain 2
Gene Aliases
ARPP
UniGene
Hs.73708
Gene Chromosome Location
Chr.10: 97572441-97583884 on Build GRCh38
Transcripts
NM_001129981.2 NM_001291218.1 NM_001291219.1 NM_020349.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3978980 c.984G>C p.R328R Chr.10: 97583626-97583626 on Build GRCh38
3868222 c.973G>A p.A325T Chr.10: 97583615-97583615 on Build GRCh38
6600454 c.1008G>A p.P336P Chr.10: 97583650-97583650 on Build GRCh38
3442427 c.951G>A p.T317T Chr.10: 97583593-97583593 on Build GRCh38
3808132 c.1051C>T p.R351* Chr.10: 97583693-97583693 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776515964 Chr.10: 97583560-97583560 on Build GRCh38
rs766898912 Chr.10: 97583530-97583530 on Build GRCh38
rs769717666 Chr.10: 97583570-97583570 on Build GRCh38
rs375635271 Chr.10: 97583629-97583629 on Build GRCh38
rs532310933 Chr.10: 97583557-97583557 on Build GRCh38
rs546297150 Chr.10: 97583499-97583499 on Build GRCh38
rs966768743 Chr.10: 97583505-97583505 on Build GRCh38
rs201529513 Chr.10: 97583665-97583665 on Build GRCh38
rs977336023 Chr.10: 97583633-97583633 on Build GRCh38
rs143138428 Chr.10: 97583578-97583578 on Build GRCh38
rs773029162 Chr.10: 97583575-97583575 on Build GRCh38
rs959833562 Chr.10: 97583545-97583545 on Build GRCh38
rs752139699 Chr.10: 97583593-97583593 on Build GRCh38
rs746233743 Chr.10: 97583549-97583549 on Build GRCh38
rs767862109 Chr.10: 97583566-97583566 on Build GRCh38
rs775418737 Chr.10: 97583605-97583605 on Build GRCh38
rs767651764 Chr.10: 97583658-97583658 on Build GRCh38
rs566640055 Chr.10: 97583625-97583625 on Build GRCh38
rs765811149 Chr.10: 97583599-97583599 on Build GRCh38
rs773157878 Chr.10: 97583650-97583650 on Build GRCh38
rs148239796 Chr.10: 97583589-97583589 on Build GRCh38
rs779481163 Chr.10: 97583547-97583547 on Build GRCh38
rs199891248 Chr.10: 97583567-97583567 on Build GRCh38
rs760242035 Chr.10: 97583674-97583674 on Build GRCh38
rs755290843 Chr.10: 97583564-97583564 on Build GRCh38
rs141723621 Chr.10: 97583613-97583613 on Build GRCh38
rs775109690 Chr.10: 97583668-97583668 on Build GRCh38
rs780442859 Chr.10: 97583555-97583555 on Build GRCh38
rs755142422 Chr.10: 97583626-97583626 on Build GRCh38
rs553884274 Chr.10: 97583529-97583529 on Build GRCh38
rs377348889 Chr.10: 97583554-97583554 on Build GRCh38
rs548309453 Chr.10: 97583590-97583590 on Build GRCh38
rs370654638 Chr.10: 97583559-97583559 on Build GRCh38
rs770719162 Chr.10: 97583654-97583654 on Build GRCh38
rs757785021 Chr.10: 97583544-97583544 on Build GRCh38
rs372700220 Chr.10: 97583649-97583649 on Build GRCh38
rs767072290 Chr.10: 97583592-97583592 on Build GRCh38
rs766115665 Chr.10: 97583586-97583586 on Build GRCh38
rs368630525 Chr.10: 97583630-97583630 on Build GRCh38
rs780214039 Chr.10: 97583624-97583624 on Build GRCh38
rs773960501 Chr.10: 97583655-97583655 on Build GRCh38
rs1040939137 Chr.10: 97583631-97583631 on Build GRCh38
rs200702482 Chr.10: 97583543-97583543 on Build GRCh38
rs749185346 Chr.10: 97583651-97583651 on Build GRCh38
rs879151949 Chr.10: 97583523-97583523 on Build GRCh38

View Full Product Details