Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12754557-12754804 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEST2
Entrez Gene ID
54831
Gene Name
bestrophin 2
Gene Aliases
VMD2L1
UniGene
Hs.435611
Gene Chromosome Location
Chr.19: 12751691-12758458 on Build GRCh38
Transcripts
NM_017682.2 XM_005259963.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
991433 c.394G>A p.A132T Chr.19: 12754698-12754698 on Build GRCh38
4493283 c.412C>A p.L138I Chr.19: 12754716-12754716 on Build GRCh38
318916 c.449G>T p.R150L Chr.19: 12754753-12754753 on Build GRCh38
5602902 c.392A>T p.Y131F Chr.19: 12754696-12754696 on Build GRCh38
991432 c.384C>A p.L128L Chr.19: 12754688-12754688 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371302296 Chr.19: 12754708-12754708 on Build GRCh38
rs754854813 Chr.19: 12754579-12754579 on Build GRCh38
rs960577644 Chr.19: 12754730-12754730 on Build GRCh38
rs199593030 Chr.19: 12754725-12754725 on Build GRCh38
rs773552289 Chr.19: 12754649-12754649 on Build GRCh38
rs754943772 Chr.19: 12754682-12754682 on Build GRCh38
rs778596981 Chr.19: 12754713-12754713 on Build GRCh38
rs566256345 Chr.19: 12754784-12754784 on Build GRCh38
rs565237088 Chr.19: 12754731-12754731 on Build GRCh38
rs541734210 Chr.19: 12754637-12754637 on Build GRCh38
rs375044336 Chr.19: 12754644-12754644 on Build GRCh38
rs758053072 Chr.19: 12754651-12754652 on Build GRCh38
rs532624266 Chr.19: 12754764-12754764 on Build GRCh38
rs773394377 Chr.19: 12754757-12754757 on Build GRCh38
rs201615329 Chr.19: 12754593-12754593 on Build GRCh38
rs759666592 Chr.19: 12754769-12754769 on Build GRCh38
rs868198303 Chr.19: 12754605-12754605 on Build GRCh38
rs745409765 Chr.19: 12754714-12754714 on Build GRCh38
rs756001142 Chr.19: 12754698-12754698 on Build GRCh38
rs767536264 Chr.19: 12754686-12754686 on Build GRCh38
rs751592629 Chr.19: 12754665-12754665 on Build GRCh38
rs747539648 Chr.19: 12754628-12754628 on Build GRCh38
rs36032297 Chr.19: 12754712-12754712 on Build GRCh38
rs369229038 Chr.19: 12754688-12754688 on Build GRCh38
rs763096626 Chr.19: 12754659-12754659 on Build GRCh38
rs771105784 Chr.19: 12754767-12754767 on Build GRCh38
rs771687530 Chr.19: 12754720-12754720 on Build GRCh38
rs865890676 Chr.19: 12754785-12754785 on Build GRCh38
rs747938909 Chr.19: 12754595-12754595 on Build GRCh38
rs748960573 Chr.19: 12754709-12754709 on Build GRCh38
rs867875103 Chr.19: 12754786-12754786 on Build GRCh38
rs1007596397 Chr.19: 12754694-12754694 on Build GRCh38
rs777534813 Chr.19: 12754615-12754615 on Build GRCh38
rs769244875 Chr.19: 12754629-12754629 on Build GRCh38
rs748886255 Chr.19: 12754616-12754616 on Build GRCh38
rs752188118 Chr.19: 12754611-12754611 on Build GRCh38
rs767485983 Chr.19: 12754772-12754772 on Build GRCh38
rs762288691 Chr.19: 12754642-12754642 on Build GRCh38
rs1050010291 Chr.19: 12754678-12754678 on Build GRCh38
rs936540167 Chr.19: 12754638-12754638 on Build GRCh38
rs368638952 Chr.19: 12754597-12754597 on Build GRCh38
rs561379374 Chr.19: 12754663-12754663 on Build GRCh38
rs777361092 Chr.19: 12754700-12754700 on Build GRCh38
rs773939596 Chr.19: 12754624-12754624 on Build GRCh38
rs34896167 Chr.19: 12754776-12754776 on Build GRCh38
rs1027845869 Chr.19: 12754685-12754685 on Build GRCh38
rs763186839 Chr.19: 12754763-12754763 on Build GRCh38
rs372694212 Chr.19: 12754626-12754626 on Build GRCh38
rs770565791 Chr.19: 12754618-12754618 on Build GRCh38

View Full Product Details