Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 70588554-70588825 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SDHAF4
Entrez Gene ID
135154
Gene Name
succinate dehydrogenase complex assembly factor 4
Gene Aliases
C6orf57, Sdh8
UniGene
Hs.418495
Gene Chromosome Location
Chr.6: 70566922-70588903 on Build GRCh38
Transcripts
NM_145267.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5007028 c.283C>T p.R95* Chr.6: 70588680-70588680 on Build GRCh38
7143204 c.250G>C p.E84Q Chr.6: 70588647-70588647 on Build GRCh38
3703047 c.229G>A p.D77N Chr.6: 70588626-70588626 on Build GRCh38
4925205 c.268G>C p.G90R Chr.6: 70588665-70588665 on Build GRCh38
4667926 c.302G>A p.R101Q Chr.6: 70588699-70588699 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762729319 Chr.6: 70588766-70588766 on Build GRCh38
rs991123463 Chr.6: 70588772-70588772 on Build GRCh38
rs759248045 Chr.6: 70588628-70588628 on Build GRCh38
rs776608369 Chr.6: 70588603-70588603 on Build GRCh38
rs777846817 Chr.6: 70588710-70588710 on Build GRCh38
rs538912127 Chr.6: 70588663-70588663 on Build GRCh38
rs776856709 Chr.6: 70588779-70588779 on Build GRCh38
rs200376688 Chr.6: 70588708-70588708 on Build GRCh38
rs767309184 Chr.6: 70588652-70588652 on Build GRCh38
rs763554671 Chr.6: 70588676-70588676 on Build GRCh38
rs148136940 Chr.6: 70588691-70588691 on Build GRCh38
rs764798487 Chr.6: 70588683-70588683 on Build GRCh38
rs771821319 Chr.6: 70588727-70588727 on Build GRCh38
rs768590845 Chr.6: 70588730-70588730 on Build GRCh38
rs942271640 Chr.6: 70588601-70588601 on Build GRCh38
rs767355667 Chr.6: 70588643-70588643 on Build GRCh38
rs749559034 Chr.6: 70588716-70588716 on Build GRCh38
rs574901961 Chr.6: 70588786-70588786 on Build GRCh38
rs1035399010 Chr.6: 70588734-70588734 on Build GRCh38
rs146446063 Chr.6: 70588620-70588620 on Build GRCh38
rs1002060986 Chr.6: 70588678-70588678 on Build GRCh38
rs756873098 Chr.6: 70588681-70588681 on Build GRCh38
rs954315015 Chr.6: 70588741-70588741 on Build GRCh38
rs142223975 Chr.6: 70588646-70588646 on Build GRCh38
rs73488105 Chr.6: 70588685-70588685 on Build GRCh38
rs572481277 Chr.6: 70588689-70588689 on Build GRCh38
rs1004161372 Chr.6: 70588693-70588693 on Build GRCh38
rs866927761 Chr.6: 70588695-70588695 on Build GRCh38
rs773962963 Chr.6: 70588604-70588604 on Build GRCh38
rs370009565 Chr.6: 70588729-70588729 on Build GRCh38
rs761222714 Chr.6: 70588746-70588746 on Build GRCh38
rs199905498 Chr.6: 70588728-70588728 on Build GRCh38
rs760562993 Chr.6: 70588699-70588699 on Build GRCh38
rs201373841 Chr.6: 70588707-70588707 on Build GRCh38
rs755569057 Chr.6: 70588698-70588698 on Build GRCh38
rs758816517 Chr.6: 70588687-70588687 on Build GRCh38
rs753306919 Chr.6: 70588680-70588680 on Build GRCh38
rs904994351 Chr.6: 70588666-70588666 on Build GRCh38
rs764858149 Chr.6: 70588755-70588755 on Build GRCh38
rs1051851755 Chr.6: 70588662-70588662 on Build GRCh38
rs184541508 Chr.6: 70588739-70588739 on Build GRCh38

View Full Product Details