Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 91281372-91281645 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC88C
Entrez Gene ID
440193
Gene Name
coiled-coil domain containing 88C
Gene Aliases
DAPLE, HKRP2, KIAA1509, SCA40
UniGene
Hs.525536
Gene Chromosome Location
Chr.14: 91271323-91418498 on Build GRCh38
Transcripts
NM_001080414.3 XM_011536796.2 XM_017021335.1 XM_017021336.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5164517 c.4674T>A p.F1558L Chr.14: 91281482-91281482 on Build GRCh38
3498723 c.4683C>T p.P1561P Chr.14: 91281473-91281473 on Build GRCh38
3498724 c.255C>T p.P85P Chr.14: 91281473-91281473 on Build GRCh38
7149383 c.247G>A p.E83K Chr.14: 91281481-91281481 on Build GRCh38
7149381 c.4675G>A p.E1559K Chr.14: 91281481-91281481 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772324109 Chr.14: 91281445-91281445 on Build GRCh38
rs1024770973 Chr.14: 91281403-91281403 on Build GRCh38
rs754306873 Chr.14: 91281426-91281426 on Build GRCh38
rs1046673645 Chr.14: 91281615-91281615 on Build GRCh38
rs747705549 Chr.14: 91281541-91281541 on Build GRCh38
rs577150883 Chr.14: 91281475-91281475 on Build GRCh38
rs948323938 Chr.14: 91281491-91281491 on Build GRCh38
rs373539289 Chr.14: 91281471-91281471 on Build GRCh38
rs139544500 Chr.14: 91281488-91281488 on Build GRCh38
rs954325136 Chr.14: 91281399-91281399 on Build GRCh38
rs779260729 Chr.14: 91281494-91281494 on Build GRCh38
rs183647972 Chr.14: 91281522-91281522 on Build GRCh38
rs947973618 Chr.14: 91281608-91281608 on Build GRCh38
rs74962163 Chr.14: 91281575-91281575 on Build GRCh38
rs181936724 Chr.14: 91281470-91281470 on Build GRCh38
rs540873605 Chr.14: 91281560-91281560 on Build GRCh38
rs544230373 Chr.14: 91281620-91281620 on Build GRCh38
rs748158814 Chr.14: 91281443-91281443 on Build GRCh38
rs755044602 Chr.14: 91281428-91281428 on Build GRCh38
rs929977146 Chr.14: 91281529-91281529 on Build GRCh38
rs75450253 Chr.14: 91281561-91281561 on Build GRCh38
rs771642816 Chr.14: 91281550-91281550 on Build GRCh38
rs773950992 Chr.14: 91281467-91281467 on Build GRCh38
rs555107791 Chr.14: 91281398-91281398 on Build GRCh38
rs747189596 Chr.14: 91281506-91281506 on Build GRCh38
rs996101869 Chr.14: 91281597-91281597 on Build GRCh38
rs749004893 Chr.14: 91281516-91281516 on Build GRCh38
rs371709826 Chr.14: 91281457-91281457 on Build GRCh38
rs778368446 Chr.14: 91281504-91281504 on Build GRCh38
rs768451252 Chr.14: 91281460-91281460 on Build GRCh38
rs923438306 Chr.14: 91281511-91281511 on Build GRCh38
rs759818807 Chr.14: 91281571-91281571 on Build GRCh38
rs537498135 Chr.14: 91281447-91281447 on Build GRCh38
rs779002989 Chr.14: 91281438-91281438 on Build GRCh38
rs775434901 Chr.14: 91281458-91281458 on Build GRCh38
rs762896355 Chr.14: 91281459-91281459 on Build GRCh38
rs779944159 Chr.14: 91281513-91281513 on Build GRCh38
rs777973415 Chr.14: 91281505-91281505 on Build GRCh38
rs201391511 Chr.14: 91281483-91281483 on Build GRCh38
rs546197796 Chr.14: 91281619-91281619 on Build GRCh38
rs761127230 Chr.14: 91281481-91281481 on Build GRCh38
rs755921000 Chr.14: 91281507-91281507 on Build GRCh38
rs774140533 Chr.14: 91281539-91281539 on Build GRCh38
rs745724134 Chr.14: 91281454-91281454 on Build GRCh38
rs376214244 Chr.14: 91281554-91281554 on Build GRCh38
rs144903315 Chr.14: 91281580-91281581 on Build GRCh38
rs530513882 Chr.14: 91281588-91281588 on Build GRCh38
rs376705157 Chr.14: 91281566-91281566 on Build GRCh38
rs752890805 Chr.14: 91281495-91281495 on Build GRCh38

View Full Product Details