Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 77988354-77988617 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP7A
Entrez Gene ID
538
Gene Name
ATPase copper transporting alpha
Gene Aliases
DSMAX, MK, MNK, SMAX3
UniGene
Hs.496414
Gene Chromosome Location
Chr.X: 77910656-78050395 on Build GRCh38
Transcripts
NM_000052.6 NM_001282224.1 NR_104109.1

SNP Detail

SNP ID Chromosome Location
rs782089301 Chr.X: 77988542-77988542 on Build GRCh38
rs371441365 Chr.X: 77988521-77988521 on Build GRCh38
rs781912401 Chr.X: 77988527-77988527 on Build GRCh38
rs782647620 Chr.X: 77988438-77988438 on Build GRCh38
rs539177302 Chr.X: 77988399-77988399 on Build GRCh38
rs797045397 Chr.X: 77988542-77988543 on Build GRCh38
rs370700841 Chr.X: 77988403-77988403 on Build GRCh38
rs782173894 Chr.X: 77988520-77988520 on Build GRCh38
rs781988009 Chr.X: 77988516-77988516 on Build GRCh38
rs937211536 Chr.X: 77988465-77988465 on Build GRCh38
rs146414063 Chr.X: 77988433-77988433 on Build GRCh38
rs781851530 Chr.X: 77988439-77988439 on Build GRCh38
rs782716555 Chr.X: 77988591-77988591 on Build GRCh38
rs368344695 Chr.X: 77988523-77988523 on Build GRCh38
rs782395208 Chr.X: 77988508-77988508 on Build GRCh38
rs782769542 Chr.X: 77988381-77988381 on Build GRCh38
rs928516350 Chr.X: 77988453-77988453 on Build GRCh38
rs61747968 Chr.X: 77988448-77988448 on Build GRCh38
rs782475540 Chr.X: 77988435-77988435 on Build GRCh38
rs61743418 Chr.X: 77988545-77988545 on Build GRCh38
rs782629239 Chr.X: 77988498-77988498 on Build GRCh38
rs782209930 Chr.X: 77988507-77988507 on Build GRCh38
rs782664232 Chr.X: 77988489-77988489 on Build GRCh38

View Full Product Details