Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 27494177-27494438 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMCO2
Entrez Gene ID
341346
Gene Name
single-pass membrane protein with coiled-coil domains 2
Gene Aliases
C12orf70
UniGene
Hs.282121
Gene Chromosome Location
Chr.12: 27446482-27506911 on Build GRCh38
Transcripts
NM_001145010.1 XM_011520634.2 XM_011520635.2 XM_011520636.2 XM_017019254.1 XM_017019255.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1586418 c.619G>T p.V207F Chr.12: 27494318-27494318 on Build GRCh38
938418 c.469G>T p.V157F Chr.12: 27494318-27494318 on Build GRCh38
938419 c.488A>C p.K163T Chr.12: 27494337-27494337 on Build GRCh38
1586416 c.638A>C p.K213T Chr.12: 27494337-27494337 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs966110195 Chr.12: 27494250-27494250 on Build GRCh38
rs898063926 Chr.12: 27494325-27494325 on Build GRCh38
rs551049816 Chr.12: 27494247-27494247 on Build GRCh38
rs930605292 Chr.12: 27494291-27494291 on Build GRCh38
rs932399504 Chr.12: 27494262-27494263 on Build GRCh38
rs937299924 Chr.12: 27494343-27494343 on Build GRCh38
rs115091099 Chr.12: 27494276-27494276 on Build GRCh38
rs538439387 Chr.12: 27494342-27494342 on Build GRCh38
rs919341317 Chr.12: 27494284-27494284 on Build GRCh38
rs763952510 Chr.12: 27494371-27494371 on Build GRCh38
rs977712826 Chr.12: 27494211-27494211 on Build GRCh38
rs761048652 Chr.12: 27494398-27494398 on Build GRCh38
rs566175612 Chr.12: 27494249-27494249 on Build GRCh38
rs894651844 Chr.12: 27494351-27494351 on Build GRCh38
rs533520629 Chr.12: 27494261-27494261 on Build GRCh38
rs549669035 Chr.12: 27494268-27494268 on Build GRCh38
rs532822095 Chr.12: 27494215-27494215 on Build GRCh38
rs1056034137 Chr.12: 27494397-27494397 on Build GRCh38
rs1012972904 Chr.12: 27494407-27494407 on Build GRCh38
rs1037128846 Chr.12: 27494318-27494318 on Build GRCh38
rs1033657817 Chr.12: 27494212-27494212 on Build GRCh38
rs1049070105 Chr.12: 27494311-27494311 on Build GRCh38
rs953682103 Chr.12: 27494216-27494216 on Build GRCh38

View Full Product Details