Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 75538049-75538320 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIP1
Entrez Gene ID
3092
Gene Name
huntingtin interacting protein 1
Gene Aliases
HIP-I, ILWEQ, SHON, SHONbeta, SHONgamma
UniGene
Hs.329266
Gene Chromosome Location
Chr.7: 75533298-75738976 on Build GRCh38
Transcripts
NM_001243198.2 NM_005338.6 XM_005250304.2 XM_005250305.2 XM_011516116.2 XM_017012099.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3882402 c.3101C>T p.T1034I Chr.7: 75538185-75538185 on Build GRCh38
5407888 c.3109G>A p.E1037K Chr.7: 75538177-75538177 on Build GRCh38
5919334 c.3076C>T p.P1026S Chr.7: 75538210-75538210 on Build GRCh38
7401245 c.3114G>C p.*1038Y Chr.7: 75538172-75538172 on Build GRCh38
3028495 c.3102C>T p.T1034T Chr.7: 75538184-75538184 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781936563 Chr.7: 75538123-75538123 on Build GRCh38
rs373051446 Chr.7: 75538236-75538236 on Build GRCh38
rs782069278 Chr.7: 75538158-75538158 on Build GRCh38
rs782670604 Chr.7: 75538237-75538237 on Build GRCh38
rs782709254 Chr.7: 75538121-75538121 on Build GRCh38
rs782185219 Chr.7: 75538262-75538262 on Build GRCh38
rs781909227 Chr.7: 75538164-75538164 on Build GRCh38
rs756756105 Chr.7: 75538083-75538083 on Build GRCh38
rs587698901 Chr.7: 75538128-75538128 on Build GRCh38
rs782186865 Chr.7: 75538209-75538209 on Build GRCh38
rs374014778 Chr.7: 75538147-75538147 on Build GRCh38
rs782542280 Chr.7: 75538228-75538228 on Build GRCh38
rs147550707 Chr.7: 75538203-75538203 on Build GRCh38
rs782084689 Chr.7: 75538144-75538144 on Build GRCh38
rs1043131199 Chr.7: 75538206-75538206 on Build GRCh38
rs782142183 Chr.7: 75538126-75538126 on Build GRCh38
rs369560175 Chr.7: 75538127-75538127 on Build GRCh38
rs1012999115 Chr.7: 75538184-75538184 on Build GRCh38
rs145040069 Chr.7: 75538072-75538073 on Build GRCh38
rs376901000 Chr.7: 75538154-75538154 on Build GRCh38
rs587754055 Chr.7: 75538180-75538180 on Build GRCh38
rs370945587 Chr.7: 75538238-75538238 on Build GRCh38
rs782510350 Chr.7: 75538175-75538175 on Build GRCh38
rs894354256 Chr.7: 75538183-75538183 on Build GRCh38
rs782480834 Chr.7: 75538191-75538191 on Build GRCh38
rs782302839 Chr.7: 75538122-75538122 on Build GRCh38
rs781817047 Chr.7: 75538181-75538181 on Build GRCh38

View Full Product Details