Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 90726618-90726859 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK14
Entrez Gene ID
5218
Gene Name
cyclin dependent kinase 14
Gene Aliases
PFTAIRE1, PFTK1
UniGene
Hs.258576
Gene Chromosome Location
Chr.7: 90595994-91210590 on Build GRCh38
Transcripts
NM_001287135.1 NM_001287136.1 NM_001287137.1 NM_012395.3 XM_005250436.3 XM_005250438.3 XM_005250439.2 XM_011516306.2 XM_011516307.2 XM_017012320.1 XM_017012321.1 XM_017012322.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
328721 c.295C>T p.R99W Chr.7: 90726792-90726792 on Build GRCh38
4671096 c.318C>T p.S106S Chr.7: 90726761-90726761 on Build GRCh38
7145076 c.369G>C p.S123S Chr.7: 90726812-90726812 on Build GRCh38
1150782 c.265C>G p.Q89E Chr.7: 90726708-90726708 on Build GRCh38
6355646 c.304A>C p.N102H Chr.7: 90726747-90726747 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762863542 Chr.7: 90726792-90726792 on Build GRCh38
rs55722725 Chr.7: 90726812-90726812 on Build GRCh38
rs760816329 Chr.7: 90726765-90726765 on Build GRCh38
rs766121316 Chr.7: 90726689-90726689 on Build GRCh38
rs747053349 Chr.7: 90726749-90726749 on Build GRCh38
rs778227720 Chr.7: 90726728-90726728 on Build GRCh38
rs775734926 Chr.7: 90726762-90726762 on Build GRCh38
rs746086232 Chr.7: 90726747-90726747 on Build GRCh38
rs773614665 Chr.7: 90726724-90726724 on Build GRCh38
rs750295045 Chr.7: 90726656-90726656 on Build GRCh38
rs773016040 Chr.7: 90726771-90726771 on Build GRCh38
rs749730201 Chr.7: 90726733-90726733 on Build GRCh38
rs921869585 Chr.7: 90726820-90726820 on Build GRCh38
rs751157761 Chr.7: 90726694-90726694 on Build GRCh38
rs756211793 Chr.7: 90726792-90726793 on Build GRCh38
rs754614156 Chr.7: 90726801-90726801 on Build GRCh38
rs780681256 Chr.7: 90726712-90726712 on Build GRCh38
rs747614328 Chr.7: 90726720-90726720 on Build GRCh38
rs367981209 Chr.7: 90726737-90726737 on Build GRCh38
rs879151175 Chr.7: 90726660-90726660 on Build GRCh38
rs138436598 Chr.7: 90726802-90726802 on Build GRCh38
rs374078260 Chr.7: 90726722-90726722 on Build GRCh38
rs187133761 Chr.7: 90726796-90726796 on Build GRCh38
rs758095992 Chr.7: 90726657-90726657 on Build GRCh38
rs771189909 Chr.7: 90726735-90726735 on Build GRCh38
rs944398673 Chr.7: 90726734-90726734 on Build GRCh38
rs548218812 Chr.7: 90726811-90726811 on Build GRCh38
rs868364699 Chr.7: 90726769-90726769 on Build GRCh38
rs765307021 Chr.7: 90726767-90726767 on Build GRCh38
rs766100371 Chr.7: 90726793-90726793 on Build GRCh38
rs754600917 Chr.7: 90726705-90726705 on Build GRCh38

View Full Product Details