Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 31126247-31126490 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INPP5J
Entrez Gene ID
27124
Gene Name
inositol polyphosphate-5-phosphatase J
Gene Aliases
INPP5, PIB5PA, PIPP
UniGene
Hs.517549
Gene Chromosome Location
Chr.22: 31107736-31134697 on Build GRCh38
Transcripts
NM_001002837.2 NM_001284285.1 NM_001284286.1 NM_001284287.1 NM_001284288.1 NM_001284289.1 XM_011530143.1 XM_017028771.1 XM_017028772.1 XM_017028773.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6686169 c.1359C>T p.S453S Chr.22: 31126463-31126463 on Build GRCh38
2937843 c.1378G>A p.A460T Chr.22: 31126482-31126482 on Build GRCh38
3553485 c.1290G>A p.W430* Chr.22: 31126394-31126394 on Build GRCh38
6686167 c.1272-8G>T p.? Chr.22: 31126368-31126368 on Build GRCh38
4692614 c.1366G>A p.A456T Chr.22: 31126470-31126470 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779032362 Chr.22: 31126326-31126326 on Build GRCh38
rs758038250 Chr.22: 31126332-31126332 on Build GRCh38
rs770642684 Chr.22: 31126345-31126345 on Build GRCh38
rs777345688 Chr.22: 31126333-31126333 on Build GRCh38
rs143502790 Chr.22: 31126447-31126447 on Build GRCh38
rs752568470 Chr.22: 31126466-31126466 on Build GRCh38
rs751134714 Chr.22: 31126421-31126421 on Build GRCh38
rs150976596 Chr.22: 31126464-31126464 on Build GRCh38
rs576889935 Chr.22: 31126397-31126397 on Build GRCh38
rs747582346 Chr.22: 31126458-31126458 on Build GRCh38
rs779254212 Chr.22: 31126448-31126448 on Build GRCh38
rs752247748 Chr.22: 31126328-31126328 on Build GRCh38
rs755252199 Chr.22: 31126347-31126347 on Build GRCh38
rs780968789 Chr.22: 31126437-31126437 on Build GRCh38
rs762781847 Chr.22: 31126467-31126467 on Build GRCh38
rs765249857 Chr.22: 31126403-31126403 on Build GRCh38
rs201357007 Chr.22: 31126449-31126449 on Build GRCh38
rs375158562 Chr.22: 31126369-31126369 on Build GRCh38
rs201915385 Chr.22: 31126391-31126391 on Build GRCh38
rs373386113 Chr.22: 31126398-31126398 on Build GRCh38
rs918318380 Chr.22: 31126455-31126455 on Build GRCh38
rs41296237 Chr.22: 31126354-31126354 on Build GRCh38
rs185711845 Chr.22: 31126469-31126469 on Build GRCh38
rs376354568 Chr.22: 31126470-31126470 on Build GRCh38
rs746572526 Chr.22: 31126335-31126335 on Build GRCh38
rs756831433 Chr.22: 31126422-31126422 on Build GRCh38
rs888655176 Chr.22: 31126304-31126304 on Build GRCh38
rs145484028 Chr.22: 31126303-31126303 on Build GRCh38
rs561823855 Chr.22: 31126463-31126463 on Build GRCh38
rs769149317 Chr.22: 31126353-31126353 on Build GRCh38
rs867947416 Chr.22: 31126312-31126312 on Build GRCh38
rs750280148 Chr.22: 31126302-31126302 on Build GRCh38
rs773142651 Chr.22: 31126457-31126457 on Build GRCh38
rs866284614 Chr.22: 31126284-31126284 on Build GRCh38
rs574415584 Chr.22: 31126349-31126349 on Build GRCh38
rs759468573 Chr.22: 31126402-31126402 on Build GRCh38
rs200683863 Chr.22: 31126452-31126452 on Build GRCh38
rs545882905 Chr.22: 31126417-31126417 on Build GRCh38
rs758356490 Chr.22: 31126412-31126412 on Build GRCh38
rs868812168 Chr.22: 31126436-31126436 on Build GRCh38
rs771979036 Chr.22: 31126374-31126374 on Build GRCh38
rs752619786 Chr.22: 31126407-31126407 on Build GRCh38
rs754506894 Chr.22: 31126400-31126400 on Build GRCh38
rs777048586 Chr.22: 31126460-31126460 on Build GRCh38

View Full Product Details