Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6916310-6916543 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENO2
Entrez Gene ID
2026
Gene Name
enolase 2
Gene Aliases
HEL-S-279, NSE
UniGene
Hs.511915
Gene Chromosome Location
Chr.12: 6914450-6923696 on Build GRCh38
Transcripts
NM_001975.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4892840 c.106C>T p.P36S Chr.12: 6916437-6916437 on Build GRCh38
6014818 c.181+8C>T p.? Chr.12: 6916520-6916520 on Build GRCh38
6654082 c.139C>A p.L47M Chr.12: 6916470-6916470 on Build GRCh38
4152941 c.129C>T p.I43I Chr.12: 6916460-6916460 on Build GRCh38
5375879 c.142G>A p.E48K Chr.12: 6916473-6916473 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782262879 Chr.12: 6916497-6916497 on Build GRCh38
rs376269766 Chr.12: 6916475-6916475 on Build GRCh38
rs929657727 Chr.12: 6916372-6916372 on Build GRCh38
rs782584259 Chr.12: 6916496-6916496 on Build GRCh38
rs781834079 Chr.12: 6916444-6916444 on Build GRCh38
rs918321807 Chr.12: 6916350-6916350 on Build GRCh38
rs782065114 Chr.12: 6916427-6916427 on Build GRCh38
rs139995547 Chr.12: 6916428-6916428 on Build GRCh38
rs782688742 Chr.12: 6916377-6916377 on Build GRCh38
rs535002011 Chr.12: 6916505-6916505 on Build GRCh38
rs782785737 Chr.12: 6916452-6916452 on Build GRCh38
rs892519952 Chr.12: 6916507-6916507 on Build GRCh38
rs202059850 Chr.12: 6916425-6916425 on Build GRCh38
rs782721628 Chr.12: 6916460-6916460 on Build GRCh38
rs199663669 Chr.12: 6916373-6916373 on Build GRCh38
rs371312536 Chr.12: 6916387-6916387 on Build GRCh38
rs374318703 Chr.12: 6916410-6916410 on Build GRCh38
rs782075941 Chr.12: 6916447-6916447 on Build GRCh38
rs781801214 Chr.12: 6916462-6916462 on Build GRCh38
rs782333945 Chr.12: 6916398-6916398 on Build GRCh38
rs781911082 Chr.12: 6916453-6916453 on Build GRCh38
rs367934778 Chr.12: 6916426-6916426 on Build GRCh38
rs11550251 Chr.12: 6916418-6916418 on Build GRCh38
rs782344801 Chr.12: 6916414-6916414 on Build GRCh38
rs782276674 Chr.12: 6916516-6916516 on Build GRCh38
rs782498815 Chr.12: 6916471-6916471 on Build GRCh38
rs782327572 Chr.12: 6916520-6916520 on Build GRCh38
rs1000634103 Chr.12: 6916457-6916457 on Build GRCh38
rs370454404 Chr.12: 6916454-6916454 on Build GRCh38
rs782583887 Chr.12: 6916403-6916403 on Build GRCh38
rs542780041 Chr.12: 6916498-6916498 on Build GRCh38
rs1031072496 Chr.12: 6916502-6916502 on Build GRCh38

View Full Product Details