Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37295760-37296033 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYTH4
Entrez Gene ID
27128
Gene Name
cytohesin 4
Gene Aliases
CYT4, DJ63G5.1, PSCD4
UniGene
Hs.170944
Gene Chromosome Location
Chr.22: 37282383-37315348 on Build GRCh38
Transcripts
NM_001318024.1 NM_013385.4 XM_011530147.2 XM_011530148.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3554136 c.174G>T p.M58I Chr.22: 37296005-37296005 on Build GRCh38
6162331 c.170G>A p.R57Q Chr.22: 37296001-37296001 on Build GRCh38
1202915 c.191A>C p.E64A Chr.22: 37296022-37296022 on Build GRCh38
726389 c.183G>A p.K61K Chr.22: 37296014-37296014 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs377622306 Chr.22: 37295968-37295968 on Build GRCh38
rs551779393 Chr.22: 37295851-37295851 on Build GRCh38
rs2269590 Chr.22: 37295866-37295866 on Build GRCh38
rs199752694 Chr.22: 37295996-37295996 on Build GRCh38
rs762933942 Chr.22: 37295983-37295990 on Build GRCh38
rs773705849 Chr.22: 37295895-37295895 on Build GRCh38
rs1041560639 Chr.22: 37295937-37295937 on Build GRCh38
rs183662549 Chr.22: 37295877-37295877 on Build GRCh38
rs777321894 Chr.22: 37295990-37295990 on Build GRCh38
rs767424515 Chr.22: 37295999-37295999 on Build GRCh38
rs187017571 Chr.22: 37295792-37295792 on Build GRCh38
rs769340898 Chr.22: 37296006-37296006 on Build GRCh38
rs369427195 Chr.22: 37296005-37296005 on Build GRCh38
rs898164731 Chr.22: 37296002-37296002 on Build GRCh38
rs531048239 Chr.22: 37295784-37295784 on Build GRCh38
rs188007002 Chr.22: 37295900-37295900 on Build GRCh38
rs753786508 Chr.22: 37295959-37295959 on Build GRCh38
rs988804517 Chr.22: 37295878-37295878 on Build GRCh38
rs972355029 Chr.22: 37295876-37295876 on Build GRCh38
rs113623649 Chr.22: 37295782-37295782 on Build GRCh38
rs970399865 Chr.22: 37295872-37295872 on Build GRCh38
rs904789653 Chr.22: 37295995-37295995 on Build GRCh38
rs754844783 Chr.22: 37295960-37295960 on Build GRCh38
rs375286906 Chr.22: 37295948-37295948 on Build GRCh38
rs367706663 Chr.22: 37295970-37295970 on Build GRCh38
rs1008263852 Chr.22: 37295800-37295800 on Build GRCh38
rs918523626 Chr.22: 37295879-37295879 on Build GRCh38
rs918051154 Chr.22: 37295880-37295880 on Build GRCh38
rs148566026 Chr.22: 37295983-37295983 on Build GRCh38
rs191761560 Chr.22: 37295867-37295867 on Build GRCh38
rs879312111 Chr.22: 37295934-37295934 on Build GRCh38
rs770573958 Chr.22: 37296000-37296000 on Build GRCh38
rs1018347624 Chr.22: 37295814-37295814 on Build GRCh38
rs942547054 Chr.22: 37295906-37295906 on Build GRCh38
rs963999907 Chr.22: 37295824-37295824 on Build GRCh38
rs771086852 Chr.22: 37296001-37296001 on Build GRCh38
rs911007074 Chr.22: 37295894-37295894 on Build GRCh38
rs530991740 Chr.22: 37295870-37295870 on Build GRCh38
rs983980616 Chr.22: 37295888-37295888 on Build GRCh38

View Full Product Details