Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67877784-67878013 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDC4
Entrez Gene ID
23644
Gene Name
enhancer of mRNA decapping 4
Gene Aliases
GE1, Ge-1, HEDL5, HEDLS, RCD-8, RCD8
UniGene
Hs.75682
Gene Chromosome Location
Chr.16: 67873023-67884514 on Build GRCh38
Transcripts
NM_014329.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6866936 c.851G>A p.S284N Chr.16: 67877802-67877802 on Build GRCh38
5148912 c.884G>A p.G295D Chr.16: 67877835-67877835 on Build GRCh38
5981003 c.887A>T p.H296L Chr.16: 67877838-67877838 on Build GRCh38
5706179 c.893C>T p.T298M Chr.16: 67877844-67877844 on Build GRCh38
4450846 c.894+1G>A p.? Chr.16: 67877846-67877846 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201977532 Chr.16: 67877852-67877852 on Build GRCh38
rs375801553 Chr.16: 67877942-67877942 on Build GRCh38
rs1027500697 Chr.16: 67877804-67877804 on Build GRCh38
rs553326569 Chr.16: 67877951-67877951 on Build GRCh38
rs757734244 Chr.16: 67877822-67877822 on Build GRCh38
rs754675491 Chr.16: 67877850-67877859 on Build GRCh38
rs770202315 Chr.16: 67877855-67877855 on Build GRCh38
rs965816819 Chr.16: 67877871-67877871 on Build GRCh38
rs780071778 Chr.16: 67877882-67877882 on Build GRCh38
rs865900738 Chr.16: 67877883-67877883 on Build GRCh38
rs751394161 Chr.16: 67877830-67877830 on Build GRCh38
rs780250156 Chr.16: 67877897-67877897 on Build GRCh38
rs946841600 Chr.16: 67877950-67877950 on Build GRCh38
rs118038841 Chr.16: 67877908-67877908 on Build GRCh38
rs564730812 Chr.16: 67877922-67877922 on Build GRCh38
rs368415512 Chr.16: 67877857-67877857 on Build GRCh38
rs922074603 Chr.16: 67877926-67877926 on Build GRCh38
rs572809321 Chr.16: 67877818-67877818 on Build GRCh38
rs373070970 Chr.16: 67877842-67877842 on Build GRCh38
rs918829701 Chr.16: 67877987-67877987 on Build GRCh38
rs370222177 Chr.16: 67877839-67877839 on Build GRCh38
rs370143766 Chr.16: 67877953-67877953 on Build GRCh38
rs8061384 Chr.16: 67877971-67877971 on Build GRCh38
rs777158469 Chr.16: 67877803-67877803 on Build GRCh38
rs950647162 Chr.16: 67877844-67877844 on Build GRCh38

View Full Product Details