Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 73953364-73953637 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COQ6
Entrez Gene ID
51004
Gene Name
coenzyme Q6, monooxygenase
Gene Aliases
CGI-10, CGI10, COQ10D6
UniGene
Hs.131555
Gene Chromosome Location
Chr.14: 73949934-73963112 on Build GRCh38
Transcripts
NM_182476.2 NM_182480.2 XM_006720156.1 XM_011536807.1 XM_011536808.1 XM_011536809.2 XM_011536810.2 XM_017021351.1 XM_017021352.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2137505 c.179T>G p.F60C Chr.14: 73953450-73953450 on Build GRCh38
2137504 c.166T>C p.Y56H Chr.14: 73953437-73953437 on Build GRCh38
6634151 c.164-10delT p.? Chr.14: 73953425-73953425 on Build GRCh38
6579296 c.229G>C p.E77Q Chr.14: 73953500-73953500 on Build GRCh38
6269696 c.278G>T p.G93V Chr.14: 73953549-73953549 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778985097 Chr.14: 73953412-73953419 on Build GRCh38
rs1035517855 Chr.14: 73953438-73953438 on Build GRCh38
rs772143356 Chr.14: 73953423-73953424 on Build GRCh38
rs751301015 Chr.14: 73953410-73953410 on Build GRCh38
rs988755478 Chr.14: 73953551-73953551 on Build GRCh38
rs771327054 Chr.14: 73953605-73953605 on Build GRCh38
rs746839544 Chr.14: 73953458-73953460 on Build GRCh38
rs979518021 Chr.14: 73953566-73953566 on Build GRCh38
rs768167170 Chr.14: 73953518-73953519 on Build GRCh38
rs191070486 Chr.14: 73953602-73953602 on Build GRCh38
rs1048374394 Chr.14: 73953603-73953603 on Build GRCh38
rs1030977081 Chr.14: 73953582-73953584 on Build GRCh38
rs138571550 Chr.14: 73953475-73953475 on Build GRCh38
rs778251454 Chr.14: 73953601-73953601 on Build GRCh38
rs774856856 Chr.14: 73953513-73953513 on Build GRCh38
rs891068944 Chr.14: 73953476-73953476 on Build GRCh38
rs558008848 Chr.14: 73953469-73953469 on Build GRCh38
rs757957335 Chr.14: 73953391-73953391 on Build GRCh38
rs781116450 Chr.14: 73953413-73953413 on Build GRCh38
rs61743864 Chr.14: 73953549-73953549 on Build GRCh38
rs774838184 Chr.14: 73953442-73953442 on Build GRCh38
rs200558787 Chr.14: 73953424-73953424 on Build GRCh38
rs755029112 Chr.14: 73953586-73953586 on Build GRCh38
rs180946215 Chr.14: 73953444-73953444 on Build GRCh38
rs751153273 Chr.14: 73953546-73953546 on Build GRCh38
rs956748438 Chr.14: 73953584-73953584 on Build GRCh38
rs574153776 Chr.14: 73953559-73953559 on Build GRCh38
rs991366555 Chr.14: 73953471-73953471 on Build GRCh38
rs772614131 Chr.14: 73953594-73953594 on Build GRCh38
rs1006718610 Chr.14: 73953539-73953539 on Build GRCh38
rs773886692 Chr.14: 73953451-73953451 on Build GRCh38
rs567714443 Chr.14: 73953431-73953431 on Build GRCh38
rs772499574 Chr.14: 73953450-73953450 on Build GRCh38
rs751908369 Chr.14: 73953437-73953437 on Build GRCh38
rs764879483 Chr.14: 73953499-73953499 on Build GRCh38
rs762722224 Chr.14: 73953536-73953536 on Build GRCh38
rs749317565 Chr.14: 73953585-73953585 on Build GRCh38
rs372647932 Chr.14: 73953558-73953558 on Build GRCh38
rs756017093 Chr.14: 73953579-73953579 on Build GRCh38
rs968368132 Chr.14: 73953516-73953516 on Build GRCh38
rs186291107 Chr.14: 73953571-73953571 on Build GRCh38
rs745746077 Chr.14: 73953419-73953420 on Build GRCh38
rs763665298 Chr.14: 73953537-73953537 on Build GRCh38
rs749567596 Chr.14: 73953425-73953425 on Build GRCh38
rs377360813 Chr.14: 73953412-73953412 on Build GRCh38
rs3213692 Chr.14: 73953424-73953424 on Build GRCh38
rs745761157 Chr.14: 73953421-73953421 on Build GRCh38
rs780249579 Chr.14: 73953583-73953583 on Build GRCh38
rs60157646 Chr.14: 73953406-73953406 on Build GRCh38
rs779008776 Chr.14: 73953588-73953588 on Build GRCh38
rs924384898 Chr.14: 73953548-73953548 on Build GRCh38
rs944156028 Chr.14: 73953573-73953573 on Build GRCh38
rs748341208 Chr.14: 73953592-73953592 on Build GRCh38

View Full Product Details