Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 11449937-11450210 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
E2F6
Entrez Gene ID
1876
Gene Name
E2F transcription factor 6
Gene Aliases
E2F-6
UniGene
Hs.603093
Gene Chromosome Location
Chr.2: 11444375-11466177 on Build GRCh38
Transcripts
NM_001278275.1 NM_001278276.1 NM_001278277.1 NM_001278278.1 NM_198256.3 NM_212540.2 NR_103490.1 XM_017003547.1 XM_017003548.1 XM_017003549.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6651036 c.590A>C p.Q197P Chr.2: 11450073-11450073 on Build GRCh38
228847 c.643C>T p.P215S Chr.2: 11450020-11450020 on Build GRCh38
6385857 c.623C>T p.T208I Chr.2: 11450040-11450040 on Build GRCh38
6418787 c.586G>T p.E196* Chr.2: 11450077-11450077 on Build GRCh38
4084343 c.579C>T p.A193A Chr.2: 11450084-11450084 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769892880 Chr.2: 11450073-11450073 on Build GRCh38
rs780096052 Chr.2: 11450146-11450146 on Build GRCh38
rs575902001 Chr.2: 11450107-11450107 on Build GRCh38
rs546723447 Chr.2: 11449972-11449972 on Build GRCh38
rs752137921 Chr.2: 11450097-11450097 on Build GRCh38
rs771450410 Chr.2: 11450079-11450079 on Build GRCh38
rs539069183 Chr.2: 11450092-11450092 on Build GRCh38
rs893552931 Chr.2: 11450069-11450069 on Build GRCh38
rs369415494 Chr.2: 11450162-11450162 on Build GRCh38
rs372597615 Chr.2: 11449965-11449965 on Build GRCh38
rs376073201 Chr.2: 11450109-11450109 on Build GRCh38
rs200951551 Chr.2: 11450160-11450160 on Build GRCh38
rs760129228 Chr.2: 11450095-11450095 on Build GRCh38
rs199506671 Chr.2: 11450096-11450096 on Build GRCh38
rs768664890 Chr.2: 11450035-11450035 on Build GRCh38
rs935102017 Chr.2: 11449992-11449992 on Build GRCh38
rs759073236 Chr.2: 11449964-11449964 on Build GRCh38
rs555220521 Chr.2: 11450182-11450182 on Build GRCh38
rs763624609 Chr.2: 11450101-11450101 on Build GRCh38
rs758327916 Chr.2: 11450126-11450126 on Build GRCh38
rs1052647878 Chr.2: 11449999-11449999 on Build GRCh38
rs745828452 Chr.2: 11450034-11450034 on Build GRCh38
rs748327105 Chr.2: 11450068-11450068 on Build GRCh38
rs189986757 Chr.2: 11450103-11450103 on Build GRCh38
rs753523023 Chr.2: 11450106-11450106 on Build GRCh38
rs778783633 Chr.2: 11450031-11450031 on Build GRCh38
rs770791215 Chr.2: 11449982-11449993 on Build GRCh38
rs371769737 Chr.2: 11450098-11450098 on Build GRCh38
rs759373860 Chr.2: 11450174-11450174 on Build GRCh38
rs752332396 Chr.2: 11449970-11449970 on Build GRCh38
rs777542849 Chr.2: 11449975-11449975 on Build GRCh38
rs527265798 Chr.2: 11450151-11450151 on Build GRCh38
rs753556695 Chr.2: 11450013-11450013 on Build GRCh38
rs781135954 Chr.2: 11450048-11450048 on Build GRCh38
rs757085346 Chr.2: 11450020-11450020 on Build GRCh38
rs141189123 Chr.2: 11450078-11450078 on Build GRCh38
rs750292850 Chr.2: 11450125-11450125 on Build GRCh38
rs774409504 Chr.2: 11450153-11450153 on Build GRCh38

View Full Product Details