Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30186708-30186941 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CORO1A
Entrez Gene ID
11151
Gene Name
coronin 1A
Gene Aliases
CLABP, CLIPINA, HCORO1, IMD8, TACO, p57
UniGene
Hs.415067
Gene Chromosome Location
Chr.16: 30183410-30189076 on Build GRCh38
Transcripts
NM_001193333.2 NM_007074.3 XM_011545714.2 XM_017022885.1 XM_017022886.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3888395 c.321G>A p.M107I Chr.16: 30186720-30186720 on Build GRCh38
3817780 c.337G>C p.D113H Chr.16: 30186831-30186831 on Build GRCh38
4819089 c.348G>C p.L116L Chr.16: 30186842-30186842 on Build GRCh38
7155989 c.328G>C p.E110Q Chr.16: 30186822-30186822 on Build GRCh38
6042964 c.436G>A p.V146M Chr.16: 30186930-30186930 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs529431628 Chr.16: 30186892-30186892 on Build GRCh38
rs775944747 Chr.16: 30186804-30186804 on Build GRCh38
rs930123430 Chr.16: 30186920-30186920 on Build GRCh38
rs756290143 Chr.16: 30186804-30186806 on Build GRCh38
rs1132812 Chr.16: 30186830-30186830 on Build GRCh38
rs767834047 Chr.16: 30186827-30186827 on Build GRCh38
rs201721540 Chr.16: 30186861-30186861 on Build GRCh38
rs760683578 Chr.16: 30186781-30186781 on Build GRCh38
rs775610075 Chr.16: 30186779-30186779 on Build GRCh38
rs1008225133 Chr.16: 30186743-30186743 on Build GRCh38
rs375095986 Chr.16: 30186799-30186799 on Build GRCh38
rs376577324 Chr.16: 30186748-30186748 on Build GRCh38
rs397514755 Chr.16: 30186894-30186894 on Build GRCh38
rs745603990 Chr.16: 30186774-30186774 on Build GRCh38
rs1007882200 Chr.16: 30186844-30186844 on Build GRCh38
rs201119953 Chr.16: 30186884-30186884 on Build GRCh38
rs566260573 Chr.16: 30186841-30186841 on Build GRCh38
rs143294920 Chr.16: 30186867-30186867 on Build GRCh38
rs370723527 Chr.16: 30186777-30186777 on Build GRCh38
rs779055659 Chr.16: 30186856-30186856 on Build GRCh38
rs755381089 Chr.16: 30186821-30186821 on Build GRCh38
rs964085070 Chr.16: 30186763-30186767 on Build GRCh38
rs200437355 Chr.16: 30186863-30186863 on Build GRCh38
rs920081881 Chr.16: 30186889-30186889 on Build GRCh38
rs764753536 Chr.16: 30186811-30186811 on Build GRCh38
rs981069144 Chr.16: 30186850-30186850 on Build GRCh38
rs545835245 Chr.16: 30186852-30186852 on Build GRCh38
rs138146826 Chr.16: 30186840-30186840 on Build GRCh38
rs761530908 Chr.16: 30186807-30186807 on Build GRCh38
rs774235337 Chr.16: 30186763-30186763 on Build GRCh38
rs370896577 Chr.16: 30186855-30186855 on Build GRCh38
rs772777006 Chr.16: 30186899-30186899 on Build GRCh38
rs770995728 Chr.16: 30186759-30186759 on Build GRCh38
rs775411755 Chr.16: 30186864-30186864 on Build GRCh38
rs149599538 Chr.16: 30186866-30186866 on Build GRCh38
rs535662377 Chr.16: 30186735-30186735 on Build GRCh38
rs756257846 Chr.16: 30186745-30186745 on Build GRCh38
rs553943700 Chr.16: 30186746-30186746 on Build GRCh38
rs1029596469 Chr.16: 30186917-30186917 on Build GRCh38
rs748628396 Chr.16: 30186736-30186736 on Build GRCh38
rs761176153 Chr.16: 30186891-30186891 on Build GRCh38
rs945998508 Chr.16: 30186728-30186728 on Build GRCh38
rs928749137 Chr.16: 30186742-30186742 on Build GRCh38
rs199732212 Chr.16: 30186812-30186812 on Build GRCh38

View Full Product Details