Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 33265014-33265245 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHMP5
Entrez Gene ID
51510
Gene Name
charged multivesicular body protein 5
Gene Aliases
C9orf83, CGI-34, HSPC177, PNAS-2, SNF7DC2, Vps60
UniGene
Hs.635313
Gene Chromosome Location
Chr.9: 33264879-33282069 on Build GRCh38
Transcripts
NM_001195536.1 NM_016410.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6245111 c.47G>T p.S16I Chr.9: 33265125-33265125 on Build GRCh38
1284117 c.1A>G p.M1V Chr.9: 33265079-33265079 on Build GRCh38
6628488 c.26A>G p.K9R Chr.9: 33265104-33265104 on Build GRCh38
6628487 c.42G>A p.P14P Chr.9: 33265120-33265120 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs373158839 Chr.9: 33265160-33265160 on Build GRCh38
rs982879209 Chr.9: 33265096-33265096 on Build GRCh38
rs752987433 Chr.9: 33265140-33265140 on Build GRCh38
rs761137293 Chr.9: 33265118-33265118 on Build GRCh38
rs779128384 Chr.9: 33265058-33265058 on Build GRCh38
rs534271250 Chr.9: 33265214-33265214 on Build GRCh38
rs748167614 Chr.9: 33265059-33265059 on Build GRCh38
rs768194877 Chr.9: 33265194-33265194 on Build GRCh38
rs745517732 Chr.9: 33265088-33265088 on Build GRCh38
rs764702824 Chr.9: 33265033-33265033 on Build GRCh38
rs754093610 Chr.9: 33265050-33265050 on Build GRCh38
rs773742450 Chr.9: 33265115-33265115 on Build GRCh38
rs6476407 Chr.9: 33265188-33265188 on Build GRCh38
rs769371689 Chr.9: 33265105-33265105 on Build GRCh38
rs952860978 Chr.9: 33265062-33265062 on Build GRCh38
rs377277774 Chr.9: 33265111-33265111 on Build GRCh38
rs765493437 Chr.9: 33265138-33265138 on Build GRCh38
rs756465540 Chr.9: 33265042-33265042 on Build GRCh38
rs755396459 Chr.9: 33265129-33265129 on Build GRCh38
rs778660606 Chr.9: 33265174-33265174 on Build GRCh38
rs770726636 Chr.9: 33265085-33265085 on Build GRCh38
rs766697807 Chr.9: 33265044-33265044 on Build GRCh38
rs906506442 Chr.9: 33265101-33265101 on Build GRCh38
rs755312569 Chr.9: 33265057-33265057 on Build GRCh38
rs781112815 Chr.9: 33265167-33265167 on Build GRCh38
rs372624153 Chr.9: 33265106-33265106 on Build GRCh38
rs762456907 Chr.9: 33265038-33265038 on Build GRCh38
rs754318300 Chr.9: 33265127-33265127 on Build GRCh38
rs780157622 Chr.9: 33265197-33265198 on Build GRCh38
rs17855469 Chr.9: 33265134-33265134 on Build GRCh38
rs769599687 Chr.9: 33265182-33265182 on Build GRCh38
rs766755613 Chr.9: 33265124-33265124 on Build GRCh38
rs981884740 Chr.9: 33265170-33265170 on Build GRCh38
rs1028766124 Chr.9: 33265173-33265173 on Build GRCh38
rs763547901 Chr.9: 33265113-33265113 on Build GRCh38
rs756184069 Chr.9: 33265188-33265189 on Build GRCh38
rs752273660 Chr.9: 33265036-33265036 on Build GRCh38
rs11540554 Chr.9: 33265128-33265128 on Build GRCh38
rs1054154057 Chr.9: 33265195-33265195 on Build GRCh38
rs751490164 Chr.9: 33265039-33265051 on Build GRCh38
rs1025004239 Chr.9: 33265047-33265047 on Build GRCh38
rs568634770 Chr.9: 33265073-33265073 on Build GRCh38
rs914905816 Chr.9: 33265215-33265215 on Build GRCh38
rs200921816 Chr.9: 33265143-33265143 on Build GRCh38
rs776456950 Chr.9: 33265087-33265087 on Build GRCh38
rs373745768 Chr.9: 33265040-33265040 on Build GRCh38
rs777754848 Chr.9: 33265147-33265147 on Build GRCh38
rs758417455 Chr.9: 33265065-33265065 on Build GRCh38
rs747980186 Chr.9: 33265186-33265186 on Build GRCh38
rs181826784 Chr.9: 33265189-33265189 on Build GRCh38

View Full Product Details