Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 72202368-72202635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDA
Entrez Gene ID
9615
Gene Name
guanine deaminase
Gene Aliases
CYPIN, GUANASE, NEDASIN
UniGene
Hs.494163
Gene Chromosome Location
Chr.9: 72114595-72256045 on Build GRCh38
Transcripts
NM_001242505.2 NM_001242506.2 NM_001242507.2 NM_004293.4 XM_005252317.1 XM_011519213.1 XM_011519214.1 XM_011519215.1 XM_011519216.1 XM_011519217.1 XM_011519218.2 XM_011519219.1 XM_011519220.1 XM_011519221.1 XM_011519222.1 XM_017015333.1 XM_017015334.1 XM_017015335.1 XM_017015336.1 XM_017015337.1 XM_017015338.1 XM_017015339.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3907909 c.276A>G p.G92G Chr.9: 72202634-72202634 on Build GRCh38
5599603 c.267C>T p.S89S Chr.9: 72202625-72202625 on Build GRCh38
1109966 c.230G>A p.G77E Chr.9: 72202588-72202588 on Build GRCh38
4432417 c.251A>G p.H84R Chr.9: 72202609-72202609 on Build GRCh38
1109964 c.219C>A p.F73L Chr.9: 72202577-72202577 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377678116 Chr.9: 72202556-72202556 on Build GRCh38
rs529545864 Chr.9: 72202604-72202604 on Build GRCh38
rs113132522 Chr.9: 72202407-72202407 on Build GRCh38
rs766896532 Chr.9: 72202581-72202581 on Build GRCh38
rs540802171 Chr.9: 72202418-72202418 on Build GRCh38
rs952725071 Chr.9: 72202511-72202511 on Build GRCh38
rs1049692446 Chr.9: 72202413-72202413 on Build GRCh38
rs1026928184 Chr.9: 72202484-72202484 on Build GRCh38
rs755331133 Chr.9: 72202610-72202610 on Build GRCh38
rs776767059 Chr.9: 72202520-72202520 on Build GRCh38
rs940410843 Chr.9: 72202445-72202445 on Build GRCh38
rs370765253 Chr.9: 72202456-72202456 on Build GRCh38
rs903318676 Chr.9: 72202412-72202412 on Build GRCh38
rs1002875117 Chr.9: 72202425-72202425 on Build GRCh38
rs1011883867 Chr.9: 72202514-72202514 on Build GRCh38
rs750486743 Chr.9: 72202557-72202557 on Build GRCh38
rs552395181 Chr.9: 72202468-72202468 on Build GRCh38
rs1023332333 Chr.9: 72202519-72202519 on Build GRCh38
rs894284449 Chr.9: 72202475-72202475 on Build GRCh38
rs199540013 Chr.9: 72202539-72202539 on Build GRCh38
rs761964345 Chr.9: 72202542-72202542 on Build GRCh38
rs886635671 Chr.9: 72202436-72202436 on Build GRCh38
rs375585337 Chr.9: 72202550-72202550 on Build GRCh38
rs1013269640 Chr.9: 72202479-72202479 on Build GRCh38
rs896256542 Chr.9: 72202467-72202467 on Build GRCh38
rs758880816 Chr.9: 72202566-72202566 on Build GRCh38
rs752037406 Chr.9: 72202592-72202592 on Build GRCh38

View Full Product Details