Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 24120987-24121212 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL22RA1
Entrez Gene ID
58985
Gene Name
interleukin 22 receptor subunit alpha 1
Gene Aliases
CRF2-9, IL22R, IL22R1
UniGene
Hs.110915
Gene Chromosome Location
Chr.1: 24119771-24143285 on Build GRCh38
Transcripts
NM_021258.3 XM_011541882.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6685236 c.1358T>C p.L453S Chr.1: 24121172-24121172 on Build GRCh38
3486373 c.1374C>T p.S458S Chr.1: 24121156-24121156 on Build GRCh38
3486372 c.1529C>T p.S510F Chr.1: 24121001-24121001 on Build GRCh38
1719146 c.1523C>T p.P508L Chr.1: 24121007-24121007 on Build GRCh38
5992604 c.1321A>G p.M441V Chr.1: 24121209-24121209 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs549934010 Chr.1: 24121068-24121068 on Build GRCh38
rs773497212 Chr.1: 24121013-24121013 on Build GRCh38
rs755538735 Chr.1: 24121057-24121057 on Build GRCh38
rs199603548 Chr.1: 24121165-24121165 on Build GRCh38
rs533465451 Chr.1: 24121168-24121168 on Build GRCh38
rs1047762708 Chr.1: 24121092-24121092 on Build GRCh38
rs907828861 Chr.1: 24121166-24121166 on Build GRCh38
rs142471602 Chr.1: 24121167-24121167 on Build GRCh38
rs137928018 Chr.1: 24121049-24121049 on Build GRCh38
rs755408667 Chr.1: 24121177-24121177 on Build GRCh38
rs780951613 Chr.1: 24121082-24121082 on Build GRCh38
rs768519535 Chr.1: 24121103-24121103 on Build GRCh38
rs761188303 Chr.1: 24121135-24121135 on Build GRCh38
rs751908237 Chr.1: 24121170-24121170 on Build GRCh38
rs764083402 Chr.1: 24121037-24121037 on Build GRCh38
rs764743291 Chr.1: 24121042-24121042 on Build GRCh38
rs769842023 Chr.1: 24121008-24121008 on Build GRCh38
rs767335066 Chr.1: 24121025-24121025 on Build GRCh38
rs781745964 Chr.1: 24121182-24121182 on Build GRCh38
rs568439273 Chr.1: 24121080-24121080 on Build GRCh38
rs951614998 Chr.1: 24121071-24121071 on Build GRCh38
rs775308000 Chr.1: 24121026-24121026 on Build GRCh38
rs752026789 Chr.1: 24121052-24121052 on Build GRCh38
rs143671796 Chr.1: 24121044-24121044 on Build GRCh38
rs369293523 Chr.1: 24121185-24121185 on Build GRCh38
rs775254539 Chr.1: 24121097-24121097 on Build GRCh38
rs760465535 Chr.1: 24121099-24121099 on Build GRCh38
rs776649354 Chr.1: 24121118-24121118 on Build GRCh38
rs769939635 Chr.1: 24121067-24121067 on Build GRCh38
rs138731413 Chr.1: 24121038-24121038 on Build GRCh38
rs760586269 Chr.1: 24121030-24121030 on Build GRCh38
rs748119811 Chr.1: 24121064-24121064 on Build GRCh38
rs771195444 Chr.1: 24121089-24121089 on Build GRCh38
rs34929192 Chr.1: 24121076-24121077 on Build GRCh38
rs529448506 Chr.1: 24121138-24121138 on Build GRCh38
rs747838224 Chr.1: 24121182-24121184 on Build GRCh38
rs756801670 Chr.1: 24121040-24121040 on Build GRCh38
rs781766039 Chr.1: 24121059-24121059 on Build GRCh38
rs1055311110 Chr.1: 24121017-24121017 on Build GRCh38
rs147491269 Chr.1: 24121039-24121039 on Build GRCh38

View Full Product Details