Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48464421-48464662 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNJ14
Entrez Gene ID
3770
Gene Name
potassium voltage-gated channel subfamily J member 14
Gene Aliases
IRK4, KIR2.4
UniGene
Hs.144011
Gene Chromosome Location
Chr.19: 48455707-48466110 on Build GRCh38
Transcripts
NM_013348.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4925090 c.977T>C p.L326P Chr.19: 48464443-48464443 on Build GRCh38
4079859 c.1039C>T p.R347C Chr.19: 48464505-48464505 on Build GRCh38
7330983 c.1077G>T p.P359P Chr.19: 48464543-48464543 on Build GRCh38
5000272 c.1039C>A p.R347S Chr.19: 48464505-48464505 on Build GRCh38
6691134 c.1114C>T p.Q372* Chr.19: 48464580-48464580 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757138715 Chr.19: 48464542-48464542 on Build GRCh38
rs201423066 Chr.19: 48464571-48464571 on Build GRCh38
rs777727987 Chr.19: 48464535-48464535 on Build GRCh38
rs562447217 Chr.19: 48464526-48464526 on Build GRCh38
rs769622863 Chr.19: 48464470-48464470 on Build GRCh38
rs35593185 Chr.19: 48464532-48464532 on Build GRCh38
rs1050799044 Chr.19: 48464562-48464562 on Build GRCh38
rs752501523 Chr.19: 48464529-48464529 on Build GRCh38
rs763573793 Chr.19: 48464561-48464561 on Build GRCh38
rs766377919 Chr.19: 48464463-48464463 on Build GRCh38
rs144271715 Chr.19: 48464559-48464559 on Build GRCh38
rs774000705 Chr.19: 48464567-48464567 on Build GRCh38
rs375438796 Chr.19: 48464498-48464498 on Build GRCh38
rs143857112 Chr.19: 48464482-48464482 on Build GRCh38
rs142079550 Chr.19: 48464582-48464582 on Build GRCh38
rs368578237 Chr.19: 48464466-48464466 on Build GRCh38
rs543602716 Chr.19: 48464598-48464598 on Build GRCh38
rs149567009 Chr.19: 48464553-48464553 on Build GRCh38
rs138120407 Chr.19: 48464543-48464543 on Build GRCh38
rs778820492 Chr.19: 48464593-48464593 on Build GRCh38
rs202154409 Chr.19: 48464499-48464499 on Build GRCh38
rs763901436 Chr.19: 48464579-48464579 on Build GRCh38
rs768869301 Chr.19: 48464483-48464483 on Build GRCh38
rs142080270 Chr.19: 48464612-48464612 on Build GRCh38
rs757193771 Chr.19: 48464585-48464585 on Build GRCh38
rs766016101 Chr.19: 48464515-48464515 on Build GRCh38
rs776868235 Chr.19: 48464492-48464492 on Build GRCh38
rs750940601 Chr.19: 48464505-48464505 on Build GRCh38
rs777018794 Chr.19: 48464572-48464572 on Build GRCh38
rs372848381 Chr.19: 48464481-48464481 on Build GRCh38
rs150484317 Chr.19: 48464537-48464537 on Build GRCh38
rs781306557 Chr.19: 48464601-48464601 on Build GRCh38
rs773723083 Chr.19: 48464631-48464631 on Build GRCh38
rs770089449 Chr.19: 48464613-48464613 on Build GRCh38
rs1054904887 Chr.19: 48464513-48464513 on Build GRCh38
rs140065287 Chr.19: 48464560-48464560 on Build GRCh38
rs562301083 Chr.19: 48464634-48464634 on Build GRCh38
rs757964679 Chr.19: 48464454-48464454 on Build GRCh38
rs201147349 Chr.19: 48464479-48464479 on Build GRCh38
rs200201274 Chr.19: 48464463-48464463 on Build GRCh38
rs146738311 Chr.19: 48464455-48464455 on Build GRCh38
rs745997313 Chr.19: 48464477-48464477 on Build GRCh38
rs747917931 Chr.19: 48464467-48464467 on Build GRCh38
rs777753341 Chr.19: 48464472-48464472 on Build GRCh38
rs754599672 Chr.19: 48464518-48464518 on Build GRCh38
rs557935092 Chr.19: 48464565-48464565 on Build GRCh38
rs139288243 Chr.19: 48464519-48464519 on Build GRCh38
rs762588558 Chr.19: 48464506-48464506 on Build GRCh38
rs867057466 Chr.19: 48464609-48464609 on Build GRCh38
rs1046922324 Chr.19: 48464520-48464520 on Build GRCh38
rs199922305 Chr.19: 48464465-48464465 on Build GRCh38
rs748506219 Chr.19: 48464557-48464557 on Build GRCh38
rs755684118 Chr.19: 48464578-48464578 on Build GRCh38
rs202102345 Chr.19: 48464517-48464517 on Build GRCh38
rs537566202 Chr.19: 48464591-48464591 on Build GRCh38

View Full Product Details