Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 22449573-22449810 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2CD5
Entrez Gene ID
9847
Gene Name
C2 calcium dependent domain containing 5
Gene Aliases
CDP138, KIAA0528
UniGene
Hs.271014
Gene Chromosome Location
Chr.12: 22448546-22544546 on Build GRCh38
Transcripts
NM_001286173.1 NM_001286174.1 NM_001286175.1 NM_001286176.1 NM_001286177.1 NM_014802.2 XM_005253538.2 XM_005253539.3 XM_006719170.2 XM_017020264.1 XM_017020265.1 XM_017020266.1 XM_017020267.1 XM_017020268.1 XM_017020269.1 XM_017020270.1 XM_017020271.1 XM_017020272.1 XM_017020273.1 XM_017020274.1 XM_017020275.1 XM_017020276.1 XM_017020277.1 XM_017020278.1 XM_017020279.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1733922 c.2989G>A p.E997K Chr.12: 22449774-22449774 on Build GRCh38
6615748 c.2532C>T p.N844N Chr.12: 22449799-22449799 on Build GRCh38
51196 c.3003+2G>A p.? Chr.12: 22449758-22449758 on Build GRCh38
1733923 c.2557G>A p.E853K Chr.12: 22449774-22449774 on Build GRCh38
6615747 c.2964C>T p.N988N Chr.12: 22449799-22449799 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs565007137 Chr.12: 22449647-22449647 on Build GRCh38
rs370776773 Chr.12: 22449788-22449788 on Build GRCh38
rs765189759 Chr.12: 22449738-22449738 on Build GRCh38
rs776223823 Chr.12: 22449718-22449718 on Build GRCh38
rs996942148 Chr.12: 22449689-22449689 on Build GRCh38
rs951862533 Chr.12: 22449701-22449701 on Build GRCh38
rs1046776 Chr.12: 22449618-22449618 on Build GRCh38
rs778491212 Chr.12: 22449751-22449751 on Build GRCh38
rs1041227866 Chr.12: 22449639-22449639 on Build GRCh38
rs754660559 Chr.12: 22449707-22449707 on Build GRCh38
rs74068581 Chr.12: 22449612-22449612 on Build GRCh38
rs549478034 Chr.12: 22449771-22449771 on Build GRCh38
rs368838207 Chr.12: 22449717-22449717 on Build GRCh38
rs1015103811 Chr.12: 22449774-22449774 on Build GRCh38
rs530737850 Chr.12: 22449688-22449688 on Build GRCh38
rs529457064 Chr.12: 22449730-22449730 on Build GRCh38
rs752483173 Chr.12: 22449764-22449764 on Build GRCh38
rs73261839 Chr.12: 22449603-22449603 on Build GRCh38
rs182360419 Chr.12: 22449713-22449713 on Build GRCh38
rs373238947 Chr.12: 22449741-22449741 on Build GRCh38
rs879406181 Chr.12: 22449771-22449772 on Build GRCh38
rs763755231 Chr.12: 22449775-22449775 on Build GRCh38
rs372491550 Chr.12: 22449693-22449693 on Build GRCh38

View Full Product Details