Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 157791630-157791869 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLINT1
Entrez Gene ID
9685
Gene Name
clathrin interactor 1
Gene Aliases
CLINT, ENTH, EPN4, EPNR
UniGene
Hs.644000
Gene Chromosome Location
Chr.5: 157785743-157859175 on Build GRCh38
Transcripts
NM_001195555.1 NM_001195556.1 NM_014666.3 XM_011534701.2 XM_017010087.1 XM_017010088.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6630327 c.1287G>A p.S429S Chr.5: 157791796-157791796 on Build GRCh38
3246993 c.1282A>C p.S428R Chr.5: 157791747-157791747 on Build GRCh38
5812857 c.1318A>T p.M440L Chr.5: 157791765-157791765 on Build GRCh38
3246995 c.1279A>G p.M427V Chr.5: 157791750-157791750 on Build GRCh38
5812858 c.1264A>T p.M422L Chr.5: 157791765-157791765 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762201040 Chr.5: 157791681-157791681 on Build GRCh38
rs755839023 Chr.5: 157791726-157791726 on Build GRCh38
rs748299730 Chr.5: 157791773-157791773 on Build GRCh38
rs763861401 Chr.5: 157791807-157791807 on Build GRCh38
rs767061476 Chr.5: 157791703-157791703 on Build GRCh38
rs768805878 Chr.5: 157791662-157791662 on Build GRCh38
rs146889751 Chr.5: 157791753-157791753 on Build GRCh38
rs763765643 Chr.5: 157791733-157791733 on Build GRCh38
rs199525692 Chr.5: 157791770-157791770 on Build GRCh38
rs758383594 Chr.5: 157791846-157791846 on Build GRCh38
rs776845410 Chr.5: 157791663-157791663 on Build GRCh38
rs755084608 Chr.5: 157791756-157791756 on Build GRCh38
rs773679234 Chr.5: 157791687-157791687 on Build GRCh38
rs995498644 Chr.5: 157791747-157791747 on Build GRCh38
rs183589548 Chr.5: 157791708-157791708 on Build GRCh38
rs757169318 Chr.5: 157791739-157791739 on Build GRCh38
rs757849845 Chr.5: 157791795-157791796 on Build GRCh38
rs917366404 Chr.5: 157791778-157791778 on Build GRCh38
rs372969322 Chr.5: 157791809-157791809 on Build GRCh38
rs759065247 Chr.5: 157791797-157791797 on Build GRCh38
rs143857615 Chr.5: 157791700-157791700 on Build GRCh38
rs751102942 Chr.5: 157791706-157791706 on Build GRCh38
rs150043675 Chr.5: 157791817-157791817 on Build GRCh38
rs770080344 Chr.5: 157791685-157791685 on Build GRCh38
rs139694588 Chr.5: 157791783-157791783 on Build GRCh38
rs767112691 Chr.5: 157791799-157791799 on Build GRCh38
rs188336524 Chr.5: 157791794-157791794 on Build GRCh38
rs778832459 Chr.5: 157791744-157791744 on Build GRCh38
rs200378943 Chr.5: 157791717-157791717 on Build GRCh38
rs753728757 Chr.5: 157791737-157791737 on Build GRCh38
rs775105603 Chr.5: 157791803-157791803 on Build GRCh38
rs200245441 Chr.5: 157791796-157791796 on Build GRCh38
rs750386491 Chr.5: 157791834-157791834 on Build GRCh38
rs753515683 Chr.5: 157791822-157791822 on Build GRCh38
rs112406734 Chr.5: 157791765-157791765 on Build GRCh38
rs765121560 Chr.5: 157791828-157791828 on Build GRCh38
rs770099070 Chr.5: 157791781-157791781 on Build GRCh38
rs1026206560 Chr.5: 157791751-157791751 on Build GRCh38
rs935989220 Chr.5: 157791831-157791831 on Build GRCh38
rs1017260743 Chr.5: 157791711-157791711 on Build GRCh38
rs757048456 Chr.5: 157791824-157791824 on Build GRCh38

View Full Product Details