Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 31336395-31336620 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSG1
Entrez Gene ID
1828
Gene Name
desmoglein 1
Gene Aliases
CDHF4, DG1, DSG, EPKHE, EPKHIA, PPKS1, SPPK1
UniGene
Hs.2633
Gene Chromosome Location
Chr.18: 31318089-31359246 on Build GRCh38
Transcripts
NM_001942.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3796362 c.1075C>A p.H359N Chr.18: 31336423-31336423 on Build GRCh38
6538894 c.1090C>G p.Q364E Chr.18: 31336438-31336438 on Build GRCh38
4140046 c.1104A>T p.K368N Chr.18: 31336452-31336452 on Build GRCh38
1630561 c.1243G>A p.G415S Chr.18: 31336591-31336591 on Build GRCh38
6538893 c.1053C>G p.V351V Chr.18: 31336401-31336401 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147377474 Chr.18: 31336566-31336566 on Build GRCh38
rs779748215 Chr.18: 31336572-31336572 on Build GRCh38
rs200411416 Chr.18: 31336556-31336556 on Build GRCh38
rs749051067 Chr.18: 31336578-31336578 on Build GRCh38
rs1003656800 Chr.18: 31336492-31336492 on Build GRCh38
rs754188744 Chr.18: 31336541-31336541 on Build GRCh38
rs776486286 Chr.18: 31336475-31336475 on Build GRCh38
rs16961655 Chr.18: 31336532-31336532 on Build GRCh38
rs752724560 Chr.18: 31336554-31336554 on Build GRCh38
rs761241711 Chr.18: 31336524-31336524 on Build GRCh38
rs755194633 Chr.18: 31336453-31336453 on Build GRCh38
rs768076202 Chr.18: 31336425-31336425 on Build GRCh38
rs376331428 Chr.18: 31336511-31336511 on Build GRCh38
rs1042409355 Chr.18: 31336433-31336433 on Build GRCh38
rs200888485 Chr.18: 31336521-31336521 on Build GRCh38
rs965717778 Chr.18: 31336499-31336499 on Build GRCh38
rs750818289 Chr.18: 31336428-31336428 on Build GRCh38
rs200495041 Chr.18: 31336504-31336504 on Build GRCh38
rs543509156 Chr.18: 31336558-31336558 on Build GRCh38
rs755179218 Chr.18: 31336542-31336542 on Build GRCh38
rs749521164 Chr.18: 31336441-31336441 on Build GRCh38
rs775246293 Chr.18: 31336498-31336498 on Build GRCh38
rs371241150 Chr.18: 31336545-31336545 on Build GRCh38
rs762555850 Chr.18: 31336502-31336502 on Build GRCh38
rs138644777 Chr.18: 31336429-31336429 on Build GRCh38
rs778888439 Chr.18: 31336456-31336456 on Build GRCh38
rs745804251 Chr.18: 31336477-31336477 on Build GRCh38
rs770787310 Chr.18: 31336466-31336466 on Build GRCh38
rs748211006 Chr.18: 31336462-31336462 on Build GRCh38
rs780551228 Chr.18: 31336439-31336439 on Build GRCh38
rs398122949 Chr.18: 31336427-31336428 on Build GRCh38
rs769638759 Chr.18: 31336488-31336488 on Build GRCh38
rs557916314 Chr.18: 31336505-31336505 on Build GRCh38
rs768303232 Chr.18: 31336503-31336503 on Build GRCh38
rs769691543 Chr.18: 31336569-31336569 on Build GRCh38
rs368707672 Chr.18: 31336555-31336555 on Build GRCh38

View Full Product Details