Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 5832428-5832653 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUT6
Entrez Gene ID
2528
Gene Name
fucosyltransferase 6
Gene Aliases
FCT3A, FT1A, Fuc-TVI, FucT-VI
UniGene
Hs.631846
Gene Chromosome Location
Chr.19: 5830626-5839753 on Build GRCh38
Transcripts
NM_000150.2 NM_001040701.1 XM_005259526.4 XM_005259527.3 XM_011527868.2 XM_011527869.2 XM_011527870.2 XM_011527872.2 XM_011527873.2 XM_011527874.2 XM_011527875.2 XM_011527876.2 XM_011527878.2 XM_011527879.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4852199 c.23A>T p.K8M Chr.19: 5832545-5832545 on Build GRCh38
7176175 c.112C>G p.Q38E Chr.19: 5832456-5832456 on Build GRCh38
3990353 c.49C>A p.L17M Chr.19: 5832519-5832519 on Build GRCh38
1207663 c.17C>T p.P6L Chr.19: 5832551-5832551 on Build GRCh38
5562406 c.122C>T p.P41L Chr.19: 5832446-5832446 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202215473 Chr.19: 5832453-5832453 on Build GRCh38
rs746409640 Chr.19: 5832528-5832528 on Build GRCh38
rs200710904 Chr.19: 5832561-5832561 on Build GRCh38
rs767265749 Chr.19: 5832488-5832488 on Build GRCh38
rs377238448 Chr.19: 5832533-5832533 on Build GRCh38
rs756805625 Chr.19: 5832526-5832526 on Build GRCh38
rs918685252 Chr.19: 5832455-5832455 on Build GRCh38
rs764810304 Chr.19: 5832596-5832596 on Build GRCh38
rs759445494 Chr.19: 5832560-5832560 on Build GRCh38
rs773317734 Chr.19: 5832557-5832557 on Build GRCh38
rs755869327 Chr.19: 5832513-5832514 on Build GRCh38
rs756682430 Chr.19: 5832594-5832594 on Build GRCh38
rs201524447 Chr.19: 5832465-5832465 on Build GRCh38
rs113764706 Chr.19: 5832505-5832505 on Build GRCh38
rs929119054 Chr.19: 5832457-5832457 on Build GRCh38
rs779718948 Chr.19: 5832589-5832589 on Build GRCh38
rs147518378 Chr.19: 5832508-5832508 on Build GRCh38
rs753219319 Chr.19: 5832521-5832521 on Build GRCh38
rs1007118895 Chr.19: 5832628-5832628 on Build GRCh38
rs371867552 Chr.19: 5832511-5832511 on Build GRCh38
rs746177959 Chr.19: 5832454-5832454 on Build GRCh38
rs368290680 Chr.19: 5832536-5832536 on Build GRCh38
rs758915187 Chr.19: 5832606-5832606 on Build GRCh38
rs766167185 Chr.19: 5832483-5832483 on Build GRCh38
rs17855737 Chr.19: 5832499-5832499 on Build GRCh38
rs568888543 Chr.19: 5832567-5832567 on Build GRCh38
rs762023607 Chr.19: 5832481-5832481 on Build GRCh38
rs762769601 Chr.19: 5832512-5832512 on Build GRCh38
rs776563221 Chr.19: 5832473-5832473 on Build GRCh38
rs751516433 Chr.19: 5832485-5832485 on Build GRCh38
rs747467061 Chr.19: 5832538-5832538 on Build GRCh38
rs144807324 Chr.19: 5832464-5832464 on Build GRCh38
rs772748812 Chr.19: 5832532-5832532 on Build GRCh38
rs775142869 Chr.19: 5832569-5832569 on Build GRCh38
rs776920409 Chr.19: 5832551-5832551 on Build GRCh38
rs763595993 Chr.19: 5832584-5832584 on Build GRCh38
rs9807877 Chr.19: 5832578-5832578 on Build GRCh38
rs749974096 Chr.19: 5832601-5832601 on Build GRCh38
rs747048069 Chr.19: 5832556-5832556 on Build GRCh38
rs753550939 Chr.19: 5832587-5832587 on Build GRCh38
rs760778966 Chr.19: 5832469-5832469 on Build GRCh38
rs374753811 Chr.19: 5832527-5832527 on Build GRCh38
rs141062404 Chr.19: 5832450-5832450 on Build GRCh38
rs143668927 Chr.19: 5832550-5832550 on Build GRCh38

View Full Product Details