Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12881135-12881402 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNASE2
Entrez Gene ID
1777
Gene Name
deoxyribonuclease II, lysosomal
Gene Aliases
DNASE2A, DNL, DNL2
UniGene
Hs.118243
Gene Chromosome Location
Chr.19: 12875211-12881521 on Build GRCh38
Transcripts
NM_001375.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1630714 c.62G>C p.G21A Chr.19: 12881314-12881314 on Build GRCh38
5031351 c.51G>A p.L17L Chr.19: 12881325-12881325 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753388409 Chr.19: 12881381-12881382 on Build GRCh38
rs766963510 Chr.19: 12881256-12881256 on Build GRCh38
rs368623153 Chr.19: 12881308-12881308 on Build GRCh38
rs755384350 Chr.19: 12881262-12881262 on Build GRCh38
rs867082577 Chr.19: 12881332-12881332 on Build GRCh38
rs1045901206 Chr.19: 12881282-12881282 on Build GRCh38
rs188695623 Chr.19: 12881192-12881192 on Build GRCh38
rs547856013 Chr.19: 12881253-12881253 on Build GRCh38
rs905895175 Chr.19: 12881208-12881208 on Build GRCh38
rs1021493478 Chr.19: 12881305-12881305 on Build GRCh38
rs867118249 Chr.19: 12881356-12881356 on Build GRCh38
rs781644094 Chr.19: 12881276-12881276 on Build GRCh38
rs891592734 Chr.19: 12881264-12881264 on Build GRCh38
rs774177992 Chr.19: 12881180-12881180 on Build GRCh38
rs897442255 Chr.19: 12881258-12881258 on Build GRCh38
rs770849055 Chr.19: 12881174-12881174 on Build GRCh38
rs574022823 Chr.19: 12881231-12881231 on Build GRCh38
rs762825662 Chr.19: 12881172-12881172 on Build GRCh38
rs758878928 Chr.19: 12881254-12881254 on Build GRCh38
rs748390311 Chr.19: 12881306-12881306 on Build GRCh38
rs745720077 Chr.19: 12881346-12881346 on Build GRCh38
rs1011224030 Chr.19: 12881361-12881361 on Build GRCh38
rs540494648 Chr.19: 12881187-12881187 on Build GRCh38
rs768011997 Chr.19: 12881270-12881271 on Build GRCh38
rs867696122 Chr.19: 12881336-12881336 on Build GRCh38
rs544404420 Chr.19: 12881274-12881274 on Build GRCh38
rs770811828 Chr.19: 12881337-12881337 on Build GRCh38
rs930205932 Chr.19: 12881281-12881281 on Build GRCh38
rs767222676 Chr.19: 12881190-12881190 on Build GRCh38
rs774263919 Chr.19: 12881342-12881342 on Build GRCh38
rs376840676 Chr.19: 12881257-12881257 on Build GRCh38
rs934094137 Chr.19: 12881188-12881188 on Build GRCh38
rs771866044 Chr.19: 12881351-12881351 on Build GRCh38
rs941321125 Chr.19: 12881217-12881217 on Build GRCh38
rs760317048 Chr.19: 12881251-12881251 on Build GRCh38
rs1010033972 Chr.19: 12881275-12881275 on Build GRCh38
rs907327839 Chr.19: 12881311-12881311 on Build GRCh38
rs911239731 Chr.19: 12881214-12881214 on Build GRCh38
rs531003217 Chr.19: 12881354-12881354 on Build GRCh38
rs1001303266 Chr.19: 12881322-12881322 on Build GRCh38
rs562700068 Chr.19: 12881329-12881329 on Build GRCh38
rs867238845 Chr.19: 12881215-12881215 on Build GRCh38
rs760275229 Chr.19: 12881368-12881368 on Build GRCh38
rs201202740 Chr.19: 12881381-12881381 on Build GRCh38

View Full Product Details