Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 48167773-48168022 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC25A
Entrez Gene ID
993
Gene Name
cell division cycle 25A
Gene Aliases
CDC25A2
UniGene
Hs.437705
Gene Chromosome Location
Chr.3: 48157178-48188658 on Build GRCh38
Transcripts
NM_001789.2 NM_201567.1 XM_006713434.2 XM_006713435.2 XM_011534316.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4644541 c.959C>A p.S320Y Chr.3: 48167916-48167916 on Build GRCh38
3916194 c.950C>T p.S317F Chr.3: 48167925-48167925 on Build GRCh38
5399517 c.1026C>T p.S342S Chr.3: 48167849-48167849 on Build GRCh38
6267747 c.956C>A p.A319E Chr.3: 48167919-48167919 on Build GRCh38
4584421 c.960T>C p.S320S Chr.3: 48167915-48167915 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs202036235 Chr.3: 48167810-48167810 on Build GRCh38
rs759544548 Chr.3: 48167987-48167987 on Build GRCh38
rs189503662 Chr.3: 48167807-48167807 on Build GRCh38
rs774368729 Chr.3: 48167986-48167986 on Build GRCh38
rs750770255 Chr.3: 48167917-48167917 on Build GRCh38
rs1025516505 Chr.3: 48167890-48167890 on Build GRCh38
rs562352342 Chr.3: 48167799-48167799 on Build GRCh38
rs769975345 Chr.3: 48167849-48167849 on Build GRCh38
rs757631724 Chr.3: 48167800-48167800 on Build GRCh38
rs377314217 Chr.3: 48167824-48167824 on Build GRCh38
rs551768306 Chr.3: 48167922-48167922 on Build GRCh38
rs953671381 Chr.3: 48167870-48167870 on Build GRCh38
rs767623713 Chr.3: 48167995-48167995 on Build GRCh38
rs552984663 Chr.3: 48167958-48167958 on Build GRCh38
rs567974501 Chr.3: 48167980-48167980 on Build GRCh38
rs776004127 Chr.3: 48167915-48167915 on Build GRCh38
rs748282795 Chr.3: 48167943-48167943 on Build GRCh38
rs767143390 Chr.3: 48167909-48167909 on Build GRCh38
rs749067218 Chr.3: 48167964-48167964 on Build GRCh38
rs373510424 Chr.3: 48167880-48167880 on Build GRCh38
rs751573758 Chr.3: 48167894-48167894 on Build GRCh38
rs138476071 Chr.3: 48167910-48167910 on Build GRCh38
rs773043129 Chr.3: 48167864-48167864 on Build GRCh38
rs35104683 Chr.3: 48167971-48167972 on Build GRCh38
rs150267735 Chr.3: 48167913-48167913 on Build GRCh38
rs747526743 Chr.3: 48167924-48167924 on Build GRCh38
rs869072226 Chr.3: 48167868-48167869 on Build GRCh38
rs374040109 Chr.3: 48167809-48167809 on Build GRCh38
rs554594251 Chr.3: 48167816-48167816 on Build GRCh38
rs569373245 Chr.3: 48167841-48167841 on Build GRCh38
rs141219026 Chr.3: 48167925-48167925 on Build GRCh38
rs1021909212 Chr.3: 48167829-48167829 on Build GRCh38
rs761449106 Chr.3: 48167918-48167918 on Build GRCh38
rs1002042589 Chr.3: 48167931-48167931 on Build GRCh38
rs755384614 Chr.3: 48167813-48167814 on Build GRCh38
rs534944126 Chr.3: 48167951-48167951 on Build GRCh38
rs766305671 Chr.3: 48167884-48167884 on Build GRCh38
rs202026442 Chr.3: 48167907-48167907 on Build GRCh38
rs780643540 Chr.3: 48167977-48167978 on Build GRCh38
rs3731539 Chr.3: 48167996-48167996 on Build GRCh38
rs776775251 Chr.3: 48167837-48167837 on Build GRCh38
rs768746187 Chr.3: 48167835-48167835 on Build GRCh38

View Full Product Details