Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 123073627-123073854 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLMP
Entrez Gene ID
79827
Gene Name
CXADR like membrane protein
Gene Aliases
ACAM, ASAM, CSBM, CSBS
UniGene
Hs.591949
Gene Chromosome Location
Chr.11: 123072006-123195305 on Build GRCh38
Transcripts
NM_024769.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6292715 c.967G>A p.A323T Chr.11: 123073629-123073629 on Build GRCh38
6630539 c.932C>T p.A311V Chr.11: 123073664-123073664 on Build GRCh38
6630538 c.946C>T p.R316W Chr.11: 123073650-123073650 on Build GRCh38
5502018 c.945G>A p.Q315Q Chr.11: 123073651-123073651 on Build GRCh38
5781563 c.943C>G p.Q315E Chr.11: 123073653-123073653 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs891090788 Chr.11: 123073670-123073670 on Build GRCh38
rs1031561696 Chr.11: 123073688-123073688 on Build GRCh38
rs375506971 Chr.11: 123073804-123073804 on Build GRCh38
rs776105463 Chr.11: 123073745-123073745 on Build GRCh38
rs906853691 Chr.11: 123073690-123073690 on Build GRCh38
rs754638208 Chr.11: 123073808-123073808 on Build GRCh38
rs760837813 Chr.11: 123073762-123073762 on Build GRCh38
rs201115327 Chr.11: 123073713-123073713 on Build GRCh38
rs758227887 Chr.11: 123073799-123073799 on Build GRCh38
rs766822478 Chr.11: 123073772-123073772 on Build GRCh38
rs375754333 Chr.11: 123073747-123073747 on Build GRCh38
rs745704648 Chr.11: 123073729-123073729 on Build GRCh38
rs889741987 Chr.11: 123073717-123073717 on Build GRCh38
rs139133124 Chr.11: 123073748-123073748 on Build GRCh38
rs749210141 Chr.11: 123073649-123073649 on Build GRCh38
rs746631557 Chr.11: 123073735-123073735 on Build GRCh38
rs151337126 Chr.11: 123073692-123073692 on Build GRCh38
rs774784052 Chr.11: 123073779-123073779 on Build GRCh38
rs763419216 Chr.11: 123073816-123073830 on Build GRCh38
rs898341255 Chr.11: 123073704-123073704 on Build GRCh38
rs765239022 Chr.11: 123073795-123073795 on Build GRCh38
rs762376722 Chr.11: 123073680-123073680 on Build GRCh38
rs371579188 Chr.11: 123073782-123073782 on Build GRCh38
rs773611572 Chr.11: 123073782-123073783 on Build GRCh38
rs773543924 Chr.11: 123073749-123073749 on Build GRCh38
rs765762289 Chr.11: 123073783-123073783 on Build GRCh38
rs978593986 Chr.11: 123073755-123073755 on Build GRCh38
rs368171739 Chr.11: 123073768-123073768 on Build GRCh38
rs770323483 Chr.11: 123073815-123073815 on Build GRCh38
rs142752755 Chr.11: 123073659-123073659 on Build GRCh38
rs370536010 Chr.11: 123073736-123073736 on Build GRCh38
rs982366113 Chr.11: 123073827-123073827 on Build GRCh38
rs112102237 Chr.11: 123073776-123073776 on Build GRCh38
rs760197866 Chr.11: 123073669-123073669 on Build GRCh38
rs777161879 Chr.11: 123073676-123073676 on Build GRCh38
rs151226212 Chr.11: 123073730-123073730 on Build GRCh38
rs766731584 Chr.11: 123073696-123073696 on Build GRCh38
rs768130579 Chr.11: 123073673-123073673 on Build GRCh38
rs139629778 Chr.11: 123073650-123073650 on Build GRCh38
rs774907281 Chr.11: 123073665-123073665 on Build GRCh38
rs767892280 Chr.11: 123073712-123073712 on Build GRCh38
rs750805043 Chr.11: 123073685-123073685 on Build GRCh38
rs765578564 Chr.11: 123073683-123073683 on Build GRCh38
rs767795254 Chr.11: 123073787-123073787 on Build GRCh38
rs920050627 Chr.11: 123073777-123073777 on Build GRCh38
rs142239120 Chr.11: 123073708-123073708 on Build GRCh38
rs377154440 Chr.11: 123073664-123073664 on Build GRCh38
rs146075432 Chr.11: 123073679-123073679 on Build GRCh38
rs750302557 Chr.11: 123073797-123073797 on Build GRCh38
rs371394874 Chr.11: 123073789-123073789 on Build GRCh38
rs775080926 Chr.11: 123073667-123073667 on Build GRCh38
rs139960670 Chr.11: 123073697-123073697 on Build GRCh38
rs781419826 Chr.11: 123073703-123073703 on Build GRCh38
rs202151595 Chr.11: 123073698-123073698 on Build GRCh38
rs778759845 Chr.11: 123073657-123073657 on Build GRCh38

View Full Product Details