Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 36844123-36844374 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1328
Entrez Gene ID
57536
Gene Name
KIAA1328
Gene Aliases
-
UniGene
Hs.4045
Gene Chromosome Location
Chr.18: 36828929-37232127 on Build GRCh38
Transcripts
NM_001322327.1 NM_020776.1 NR_136303.1 NR_136304.1 NR_136305.1 XM_005258315.4 XM_005258317.3 XM_005258318.4 XM_011526099.2 XM_011526100.2 XM_017025876.1 XM_017025877.1 XM_017025878.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6990307 c.313T>G p.L105V Chr.18: 36844283-36844283 on Build GRCh38
988256 c.329C>T p.A110V Chr.18: 36844299-36844299 on Build GRCh38
4275049 c.258A>C p.S86S Chr.18: 36844240-36844240 on Build GRCh38
5005449 c.290G>A p.R97H Chr.18: 36844272-36844272 on Build GRCh38
6298298 c.238G>T p.G80* Chr.18: 36844220-36844220 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773495684 Chr.18: 36844328-36844328 on Build GRCh38
rs762133917 Chr.18: 36844204-36844204 on Build GRCh38
rs750806827 Chr.18: 36844212-36844212 on Build GRCh38
rs775366555 Chr.18: 36844158-36844158 on Build GRCh38
rs554212352 Chr.18: 36844192-36844192 on Build GRCh38
rs767737590 Chr.18: 36844306-36844306 on Build GRCh38
rs1014912081 Chr.18: 36844315-36844315 on Build GRCh38
rs777655826 Chr.18: 36844264-36844264 on Build GRCh38
rs764210006 Chr.18: 36844186-36844186 on Build GRCh38
rs754570251 Chr.18: 36844236-36844236 on Build GRCh38
rs756518965 Chr.18: 36844216-36844216 on Build GRCh38
rs768820620 Chr.18: 36844288-36844288 on Build GRCh38
rs960784872 Chr.18: 36844316-36844316 on Build GRCh38
rs766326920 Chr.18: 36844192-36844193 on Build GRCh38
rs371261522 Chr.18: 36844182-36844182 on Build GRCh38
rs746827088 Chr.18: 36844271-36844271 on Build GRCh38
rs763159873 Chr.18: 36844159-36844159 on Build GRCh38
rs372565622 Chr.18: 36844193-36844193 on Build GRCh38
rs373756824 Chr.18: 36844298-36844298 on Build GRCh38
rs778680947 Chr.18: 36844246-36844246 on Build GRCh38
rs752853218 Chr.18: 36844308-36844309 on Build GRCh38
rs751827847 Chr.18: 36844195-36844195 on Build GRCh38
rs747766336 Chr.18: 36844252-36844252 on Build GRCh38
rs757230425 Chr.18: 36844156-36844156 on Build GRCh38
rs1040495909 Chr.18: 36844215-36844215 on Build GRCh38
rs774445519 Chr.18: 36844290-36844290 on Build GRCh38
rs753280350 Chr.18: 36844235-36844235 on Build GRCh38
rs770771022 Chr.18: 36844274-36844274 on Build GRCh38
rs780613173 Chr.18: 36844234-36844234 on Build GRCh38
rs376746767 Chr.18: 36844314-36844314 on Build GRCh38
rs776689055 Chr.18: 36844275-36844275 on Build GRCh38
rs767827727 Chr.18: 36844207-36844207 on Build GRCh38
rs903289326 Chr.18: 36844255-36844255 on Build GRCh38
rs771876001 Chr.18: 36844253-36844253 on Build GRCh38
rs761165047 Chr.18: 36844334-36844334 on Build GRCh38
rs762919678 Chr.18: 36844279-36844279 on Build GRCh38
rs766777762 Chr.18: 36844342-36844342 on Build GRCh38
rs765444801 Chr.18: 36844306-36844307 on Build GRCh38
rs777919461 Chr.18: 36844184-36844184 on Build GRCh38
rs759621392 Chr.18: 36844283-36844286 on Build GRCh38

View Full Product Details