Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 123615078-123615351 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX55
Entrez Gene ID
57696
Gene Name
DEAD-box helicase 55
Gene Aliases
-
UniGene
Hs.286173
Gene Chromosome Location
Chr.12: 123602077-123620943 on Build GRCh38
Transcripts
NM_020936.2 NR_135104.1 NR_135105.1 XM_011538608.2 XM_017019710.1 XM_017019711.1 XM_017019712.1 XM_017019713.1 XM_017019714.1 XM_017019715.1 XM_017019716.1 XM_017019717.1 XM_017019718.1 XM_017019719.1 XM_017019720.1 XM_017019721.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4676880 c.940T>C p.F314L Chr.12: 123615300-123615300 on Build GRCh38
6895014 c.936G>T p.M312I Chr.12: 123615296-123615296 on Build GRCh38
236086 c.933C>A p.F311L Chr.12: 123615293-123615293 on Build GRCh38
7203487 c.917A>C p.K306T Chr.12: 123615277-123615277 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs538166521 Chr.12: 123615129-123615129 on Build GRCh38
rs773816120 Chr.12: 123615323-123615323 on Build GRCh38
rs773084082 Chr.12: 123615213-123615213 on Build GRCh38
rs754627470 Chr.12: 123615265-123615265 on Build GRCh38
rs771794295 Chr.12: 123615292-123615292 on Build GRCh38
rs753206652 Chr.12: 123615161-123615161 on Build GRCh38
rs1049524248 Chr.12: 123615322-123615322 on Build GRCh38
rs748903983 Chr.12: 123615197-123615197 on Build GRCh38
rs542484971 Chr.12: 123615165-123615165 on Build GRCh38
rs992501287 Chr.12: 123615120-123615120 on Build GRCh38
rs934608349 Chr.12: 123615230-123615230 on Build GRCh38
rs774172727 Chr.12: 123615203-123615203 on Build GRCh38
rs752057408 Chr.12: 123615257-123615257 on Build GRCh38
rs776754053 Chr.12: 123615308-123615308 on Build GRCh38
rs779612292 Chr.12: 123615261-123615261 on Build GRCh38
rs771054400 Chr.12: 123615303-123615303 on Build GRCh38
rs778449815 Chr.12: 123615252-123615252 on Build GRCh38
rs78864418 Chr.12: 123615155-123615155 on Build GRCh38
rs773135451 Chr.12: 123615294-123615294 on Build GRCh38
rs753078772 Chr.12: 123615240-123615240 on Build GRCh38
rs758798320 Chr.12: 123615251-123615251 on Build GRCh38
rs761696936 Chr.12: 123615207-123615207 on Build GRCh38
rs35708602 Chr.12: 123615157-123615157 on Build GRCh38
rs770445047 Chr.12: 123615207-123615208 on Build GRCh38
rs929894768 Chr.12: 123615130-123615130 on Build GRCh38
rs114627783 Chr.12: 123615166-123615166 on Build GRCh38
rs1016579627 Chr.12: 123615107-123615107 on Build GRCh38
rs546194499 Chr.12: 123615280-123615280 on Build GRCh38
rs772027304 Chr.12: 123615212-123615212 on Build GRCh38
rs779588250 Chr.12: 123615188-123615188 on Build GRCh38
rs768403853 Chr.12: 123615201-123615201 on Build GRCh38
rs1035630449 Chr.12: 123615132-123615132 on Build GRCh38
rs766377668 Chr.12: 123615229-123615229 on Build GRCh38
rs765572786 Chr.12: 123615239-123615239 on Build GRCh38
rs139308305 Chr.12: 123615185-123615185 on Build GRCh38
rs745405419 Chr.12: 123615168-123615168 on Build GRCh38
rs759787513 Chr.12: 123615236-123615236 on Build GRCh38
rs17852729 Chr.12: 123615215-123615215 on Build GRCh38
rs371091632 Chr.12: 123615295-123615295 on Build GRCh38
rs11612770 Chr.12: 123615260-123615260 on Build GRCh38
rs934093413 Chr.12: 123615206-123615206 on Build GRCh38
rs748627442 Chr.12: 123615205-123615206 on Build GRCh38
rs984023352 Chr.12: 123615148-123615148 on Build GRCh38
rs754070545 Chr.12: 123615231-123615231 on Build GRCh38
rs778387510 Chr.12: 123615279-123615279 on Build GRCh38
rs769381487 Chr.12: 123615187-123615187 on Build GRCh38

View Full Product Details