Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 149752383-149752626 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COMMD2
Entrez Gene ID
51122
Gene Name
COMM domain containing 2
Gene Aliases
HSPC042
UniGene
Hs.432729
Gene Chromosome Location
Chr.3: 149737160-149752507 on Build GRCh38
Transcripts
NM_016094.3 NR_135149.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5580691 c.45C>T p.F15F Chr.3: 149752400-149752400 on Build GRCh38
4584030 c.24G>A p.E8E Chr.3: 149752421-149752421 on Build GRCh38
1752962 c.19G>A p.E7K Chr.3: 149752426-149752426 on Build GRCh38
5773580 c.8T>A p.L3Q Chr.3: 149752437-149752437 on Build GRCh38
6011239 c.42C>T p.A14A Chr.3: 149752403-149752403 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753575596 Chr.3: 149752427-149752427 on Build GRCh38
rs777401512 Chr.3: 149752468-149752468 on Build GRCh38
rs375933323 Chr.3: 149752454-149752454 on Build GRCh38
rs774245187 Chr.3: 149752429-149752429 on Build GRCh38
rs565018495 Chr.3: 149752484-149752484 on Build GRCh38
rs1009168065 Chr.3: 149752576-149752576 on Build GRCh38
rs377353632 Chr.3: 149752433-149752433 on Build GRCh38
rs374397975 Chr.3: 149752493-149752493 on Build GRCh38
rs368764382 Chr.3: 149752481-149752481 on Build GRCh38
rs763789500 Chr.3: 149752422-149752422 on Build GRCh38
rs941190234 Chr.3: 149752501-149752501 on Build GRCh38
rs368706725 Chr.3: 149752475-149752475 on Build GRCh38
rs992649966 Chr.3: 149752404-149752404 on Build GRCh38
rs933941508 Chr.3: 149752539-149752539 on Build GRCh38
rs138597206 Chr.3: 149752461-149752461 on Build GRCh38
rs767299132 Chr.3: 149752434-149752434 on Build GRCh38
rs1039523927 Chr.3: 149752504-149752504 on Build GRCh38
rs764206234 Chr.3: 149752450-149752450 on Build GRCh38
rs200287559 Chr.3: 149752452-149752452 on Build GRCh38
rs745747875 Chr.3: 149752476-149752476 on Build GRCh38
rs1001231191 Chr.3: 149752513-149752513 on Build GRCh38
rs551496239 Chr.3: 149752478-149752478 on Build GRCh38
rs899751767 Chr.3: 149752521-149752521 on Build GRCh38
rs903583334 Chr.3: 149752603-149752603 on Build GRCh38
rs917275460 Chr.3: 149752480-149752480 on Build GRCh38
rs753140224 Chr.3: 149752406-149752407 on Build GRCh38
rs1020038314 Chr.3: 149752588-149752588 on Build GRCh38
rs765892629 Chr.3: 149752458-149752458 on Build GRCh38
rs776970730 Chr.3: 149752487-149752487 on Build GRCh38
rs750910593 Chr.3: 149752459-149752459 on Build GRCh38
rs769193180 Chr.3: 149752415-149752415 on Build GRCh38
rs1022018382 Chr.3: 149752581-149752581 on Build GRCh38
rs373013910 Chr.3: 149752457-149752457 on Build GRCh38
rs760890306 Chr.3: 149752443-149752443 on Build GRCh38
rs762006297 Chr.3: 149752490-149752490 on Build GRCh38
rs543028868 Chr.3: 149752408-149752408 on Build GRCh38
rs909947413 Chr.3: 149752494-149752494 on Build GRCh38
rs371002136 Chr.3: 149752469-149752469 on Build GRCh38
rs772723007 Chr.3: 149752421-149752421 on Build GRCh38
rs775575144 Chr.3: 149752440-149752440 on Build GRCh38
rs1014040455 Chr.3: 149752585-149752585 on Build GRCh38
rs747797574 Chr.3: 149752409-149752409 on Build GRCh38
rs765684940 Chr.3: 149752556-149752556 on Build GRCh38
rs1403027 Chr.3: 149752467-149752467 on Build GRCh38
rs756488908 Chr.3: 149752463-149752464 on Build GRCh38
rs778098519 Chr.3: 149752465-149752466 on Build GRCh38
rs564057021 Chr.3: 149752451-149752451 on Build GRCh38
rs550689958 Chr.3: 149752558-149752558 on Build GRCh38
rs371183639 Chr.3: 149752485-149752485 on Build GRCh38
rs375475203 Chr.3: 149752473-149752473 on Build GRCh38
rs892153551 Chr.3: 149752563-149752563 on Build GRCh38

View Full Product Details