Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 11973774-11974033 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEFB136
Entrez Gene ID
613210
Gene Name
defensin beta 136
Gene Aliases
DEFB137
UniGene
Hs.558285
Gene Chromosome Location
Chr.8: 11973937-11974599 on Build GRCh38
Transcripts
NM_001033018.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1177027 c.196C>T p.R66C Chr.8: 11973978-11973978 on Build GRCh38
1203324 c.187A>G p.N63D Chr.8: 11973987-11973987 on Build GRCh38
7210133 c.210G>A p.P70P Chr.8: 11973964-11973964 on Build GRCh38
6642211 c.209_210insT p.Q71fs*>9 Chr.8: 11973964-11973965 on Build GRCh38
3644536 c.232C>T p.H78Y Chr.8: 11973942-11973942 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373016932 Chr.8: 11973990-11973990 on Build GRCh38
rs766007942 Chr.8: 11973933-11973933 on Build GRCh38
rs746213153 Chr.8: 11973901-11973901 on Build GRCh38
rs746967854 Chr.8: 11973896-11973896 on Build GRCh38
rs749384025 Chr.8: 11973904-11973904 on Build GRCh38
rs199745025 Chr.8: 11973968-11973968 on Build GRCh38
rs947999268 Chr.8: 11973874-11973874 on Build GRCh38
rs774225070 Chr.8: 11973918-11973918 on Build GRCh38
rs776362614 Chr.8: 11973934-11973934 on Build GRCh38
rs530122364 Chr.8: 11973890-11973890 on Build GRCh38
rs921052281 Chr.8: 11973980-11973980 on Build GRCh38
rs887391053 Chr.8: 11973808-11973808 on Build GRCh38
rs376471850 Chr.8: 11974001-11974001 on Build GRCh38
rs759045946 Chr.8: 11973936-11973936 on Build GRCh38
rs776251811 Chr.8: 11973994-11973994 on Build GRCh38
rs372958569 Chr.8: 11973943-11973943 on Build GRCh38
rs918350911 Chr.8: 11973841-11973841 on Build GRCh38
rs755111812 Chr.8: 11973965-11973965 on Build GRCh38
rs201142786 Chr.8: 11974005-11974005 on Build GRCh38
rs972441869 Chr.8: 11973970-11973970 on Build GRCh38
rs780222605 Chr.8: 11973963-11973963 on Build GRCh38
rs771980272 Chr.8: 11973920-11973920 on Build GRCh38
rs753230784 Chr.8: 11973909-11973909 on Build GRCh38
rs371702520 Chr.8: 11973886-11973886 on Build GRCh38
rs780317062 Chr.8: 11973902-11973902 on Build GRCh38
rs748199298 Chr.8: 11973977-11973977 on Build GRCh38
rs754092314 Chr.8: 11973964-11973964 on Build GRCh38
rs939141779 Chr.8: 11973845-11973845 on Build GRCh38
rs376444829 Chr.8: 11973893-11973893 on Build GRCh38
rs529064914 Chr.8: 11973875-11973875 on Build GRCh38
rs182273296 Chr.8: 11973938-11973938 on Build GRCh38
rs756532390 Chr.8: 11973948-11973948 on Build GRCh38
rs772068307 Chr.8: 11973978-11973978 on Build GRCh38
rs752206698 Chr.8: 11973941-11973941 on Build GRCh38
rs991994852 Chr.8: 11973863-11973863 on Build GRCh38
rs752112053 Chr.8: 11973894-11973894 on Build GRCh38
rs940285273 Chr.8: 11973821-11973821 on Build GRCh38
rs369065136 Chr.8: 11973929-11973929 on Build GRCh38
rs376086722 Chr.8: 11973945-11973945 on Build GRCh38
rs147613544 Chr.8: 11973914-11973915 on Build GRCh38
rs1049109552 Chr.8: 11973804-11973804 on Build GRCh38
rs757799327 Chr.8: 11973895-11973895 on Build GRCh38
rs569033424 Chr.8: 11973897-11973897 on Build GRCh38
rs988070055 Chr.8: 11973966-11973966 on Build GRCh38
rs796365227 Chr.8: 11973862-11973862 on Build GRCh38
rs748111302 Chr.8: 11973919-11973919 on Build GRCh38
rs368241297 Chr.8: 11973829-11973829 on Build GRCh38
rs916456378 Chr.8: 11974006-11974006 on Build GRCh38
rs746893199 Chr.8: 11973986-11973986 on Build GRCh38
rs189995324 Chr.8: 11973923-11973923 on Build GRCh38
rs916466540 Chr.8: 11973870-11973870 on Build GRCh38
rs895830799 Chr.8: 11973831-11973831 on Build GRCh38
rs533233153 Chr.8: 11973914-11973914 on Build GRCh38

View Full Product Details