Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 32030939-32031186 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA7
Entrez Gene ID
1139
Gene Name
cholinergic receptor nicotinic alpha 7 subunit
Gene Aliases
CHRNA7-2, NACHRA7
UniGene
Hs.511772
Gene Chromosome Location
Chr.15: 32030462-32172521 on Build GRCh38
Transcripts
NM_000746.5 NM_001190455.2 NR_046324.1 XM_011521178.2 XM_017021882.1 XM_017021883.1 XM_017021884.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1660241 c.111delC p.Y37fs*1 Chr.15: 32030953-32030953 on Build GRCh38
4250297 c.282+10G>A p.? Chr.15: 32031047-32031047 on Build GRCh38
7202950 c.124A>C p.R42R Chr.15: 32030966-32030966 on Build GRCh38
1248294 c.157G>A p.V53I Chr.15: 32030999-32030999 on Build GRCh38
7202949 c.211A>C p.R71R Chr.15: 32030966-32030966 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199974614 Chr.15: 32030998-32030998 on Build GRCh38
rs887938407 Chr.15: 32031066-32031066 on Build GRCh38
rs759217449 Chr.15: 32031065-32031065 on Build GRCh38
rs1019843752 Chr.15: 32030968-32030968 on Build GRCh38
rs75349629 Chr.15: 32031070-32031070 on Build GRCh38
rs910602916 Chr.15: 32031157-32031157 on Build GRCh38
rs199509689 Chr.15: 32031075-32031075 on Build GRCh38
rs369310804 Chr.15: 32031037-32031037 on Build GRCh38
rs770216397 Chr.15: 32031086-32031086 on Build GRCh38
rs1019897922 Chr.15: 32031162-32031162 on Build GRCh38
rs752564344 Chr.15: 32030992-32030992 on Build GRCh38
rs201301097 Chr.15: 32031072-32031072 on Build GRCh38
rs949020285 Chr.15: 32031090-32031090 on Build GRCh38
rs201348759 Chr.15: 32031057-32031057 on Build GRCh38
rs769072651 Chr.15: 32030971-32030971 on Build GRCh38
rs1047128004 Chr.15: 32031140-32031140 on Build GRCh38
rs372738902 Chr.15: 32031108-32031108 on Build GRCh38
rs561821235 Chr.15: 32031116-32031116 on Build GRCh38
rs752513276 Chr.15: 32031071-32031071 on Build GRCh38
rs968278669 Chr.15: 32031149-32031149 on Build GRCh38
rs981995937 Chr.15: 32031104-32031104 on Build GRCh38
rs887114198 Chr.15: 32031143-32031143 on Build GRCh38
rs200783326 Chr.15: 32031085-32031085 on Build GRCh38
rs898598423 Chr.15: 32031121-32031121 on Build GRCh38
rs777062479 Chr.15: 32031055-32031055 on Build GRCh38
rs553156178 Chr.15: 32031042-32031042 on Build GRCh38
rs202232041 Chr.15: 32031035-32031035 on Build GRCh38
rs746027896 Chr.15: 32031023-32031023 on Build GRCh38
rs1032521091 Chr.15: 32031020-32031020 on Build GRCh38
rs780664892 Chr.15: 32030967-32030967 on Build GRCh38
rs202124963 Chr.15: 32031059-32031059 on Build GRCh38
rs368352998 Chr.15: 32030985-32030985 on Build GRCh38
rs376276457 Chr.15: 32031052-32031052 on Build GRCh38
rs148523647 Chr.15: 32031112-32031112 on Build GRCh38
rs772745507 Chr.15: 32030978-32030978 on Build GRCh38
rs372054567 Chr.15: 32031003-32031003 on Build GRCh38
rs201094833 Chr.15: 32031024-32031024 on Build GRCh38
rs778912338 Chr.15: 32031088-32031088 on Build GRCh38
rs910672291 Chr.15: 32030989-32030989 on Build GRCh38
rs551823254 Chr.15: 32031028-32031028 on Build GRCh38
rs564494012 Chr.15: 32031152-32031152 on Build GRCh38
rs766039837 Chr.15: 32030987-32030987 on Build GRCh38
rs747413506 Chr.15: 32031051-32031051 on Build GRCh38
rs116271692 Chr.15: 32031124-32031124 on Build GRCh38
rs201529515 Chr.15: 32030979-32030979 on Build GRCh38
rs200780407 Chr.15: 32030993-32030993 on Build GRCh38
rs544312066 Chr.15: 32031139-32031139 on Build GRCh38
rs763428797 Chr.15: 32031130-32031130 on Build GRCh38
rs148052103 Chr.15: 32030995-32030995 on Build GRCh38
rs757343352 Chr.15: 32030999-32030999 on Build GRCh38
rs767364343 Chr.15: 32030990-32030990 on Build GRCh38
rs994918783 Chr.15: 32031119-32031119 on Build GRCh38
rs200147286 Chr.15: 32030988-32030988 on Build GRCh38
rs200234247 Chr.15: 32031045-32031045 on Build GRCh38
rs943445847 Chr.15: 32031167-32031167 on Build GRCh38
rs760440514 Chr.15: 32031077-32031077 on Build GRCh38
rs34688612 Chr.15: 32030965-32030965 on Build GRCh38
rs764082164 Chr.15: 32031081-32031081 on Build GRCh38

View Full Product Details