Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 136242780-136243035 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTFR2
Entrez Gene ID
113115
Gene Name
mitochondrial fission regulator 2
Gene Aliases
DUFD1, FAM54A
UniGene
Hs.121536
Gene Chromosome Location
Chr.6: 136231030-136253659 on Build GRCh38
Transcripts
NM_001099286.2 NM_001318738.1 NM_138419.4 XM_011535410.2 XM_011535412.2 XM_011535413.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5884308 c.280C>T p.R94* Chr.6: 136242862-136242862 on Build GRCh38
1440646 c.225T>G p.S75S Chr.6: 136242917-136242917 on Build GRCh38
5557496 c.235A>G p.I79V Chr.6: 136242907-136242907 on Build GRCh38
4920047 c.196T>C p.S66P Chr.6: 136242946-136242946 on Build GRCh38
1073664 c.177G>T p.P59P Chr.6: 136242965-136242965 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs866249054 Chr.6: 136243013-136243013 on Build GRCh38
rs988723624 Chr.6: 136242835-136242835 on Build GRCh38
rs766309585 Chr.6: 136242980-136242980 on Build GRCh38
rs201167393 Chr.6: 136242862-136242862 on Build GRCh38
rs751278670 Chr.6: 136242880-136242880 on Build GRCh38
rs775647329 Chr.6: 136242923-136242923 on Build GRCh38
rs368248303 Chr.6: 136242828-136242828 on Build GRCh38
rs139025577 Chr.6: 136242942-136242942 on Build GRCh38
rs151189804 Chr.6: 136242817-136242817 on Build GRCh38
rs761864764 Chr.6: 136242945-136242945 on Build GRCh38
rs768557196 Chr.6: 136242939-136242939 on Build GRCh38
rs369199068 Chr.6: 136242839-136242839 on Build GRCh38
rs747256886 Chr.6: 136242928-136242928 on Build GRCh38
rs570529220 Chr.6: 136242966-136242966 on Build GRCh38
rs767134117 Chr.6: 136243002-136243002 on Build GRCh38
rs190085569 Chr.6: 136242861-136242861 on Build GRCh38
rs757769713 Chr.6: 136242898-136242898 on Build GRCh38
rs762786528 Chr.6: 136242975-136242975 on Build GRCh38
rs772478044 Chr.6: 136242909-136242909 on Build GRCh38
rs771134744 Chr.6: 136242810-136242810 on Build GRCh38
rs756252114 Chr.6: 136242949-136242949 on Build GRCh38
rs140325976 Chr.6: 136243014-136243014 on Build GRCh38
rs767946861 Chr.6: 136242944-136242945 on Build GRCh38
rs750815255 Chr.6: 136242989-136242989 on Build GRCh38
rs777834176 Chr.6: 136242976-136242976 on Build GRCh38
rs973829785 Chr.6: 136242902-136242902 on Build GRCh38
rs756388356 Chr.6: 136242997-136243000 on Build GRCh38
rs773330439 Chr.6: 136242972-136242972 on Build GRCh38
rs759240326 Chr.6: 136242986-136242986 on Build GRCh38
rs146917381 Chr.6: 136242896-136242896 on Build GRCh38
rs114903986 Chr.6: 136242841-136242841 on Build GRCh38
rs374864482 Chr.6: 136242831-136242831 on Build GRCh38
rs764262780 Chr.6: 136242830-136242830 on Build GRCh38
rs754407639 Chr.6: 136242887-136242887 on Build GRCh38
rs949202097 Chr.6: 136242872-136242872 on Build GRCh38
rs774712767 Chr.6: 136242811-136242811 on Build GRCh38
rs746204271 Chr.6: 136242906-136242906 on Build GRCh38
rs763716888 Chr.6: 136242826-136242826 on Build GRCh38
rs757757729 Chr.6: 136242906-136242907 on Build GRCh38
rs905997503 Chr.6: 136242815-136242815 on Build GRCh38
rs368063262 Chr.6: 136242827-136242827 on Build GRCh38
rs916424969 Chr.6: 136242843-136242843 on Build GRCh38
rs779466385 Chr.6: 136242904-136242904 on Build GRCh38
rs780760418 Chr.6: 136242889-136242889 on Build GRCh38

View Full Product Details