Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 63721380-63721637 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EYS
Entrez Gene ID
346007
Gene Name
eyes shut homolog (Drosophila)
Gene Aliases
C6orf178, C6orf179, C6orf180, EGFL10, EGFL11, RP25, SPAM, bA166P24.2, bA307F22.3, bA74E24.1, dJ1018A4.2, dJ22I17.2, dJ303F19.1
UniGene
Hs.25067
Gene Chromosome Location
Chr.6: 63719980-65707225 on Build GRCh38
Transcripts
NM_001142800.1 NM_001292009.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6888902 c.8654G>A p.G2885E Chr.6: 63721440-63721440 on Build GRCh38
4824359 c.8620A>C p.I2874L Chr.6: 63721474-63721474 on Build GRCh38
3830929 c.8499G>T p.M2833I Chr.6: 63721595-63721595 on Build GRCh38
1445560 c.8593T>G p.F2865V Chr.6: 63721501-63721501 on Build GRCh38
3394272 c.8536G>A p.G2846R Chr.6: 63721558-63721558 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746440557 Chr.6: 63721404-63721404 on Build GRCh38
rs1009311910 Chr.6: 63721574-63721574 on Build GRCh38
rs1056065852 Chr.6: 63721536-63721536 on Build GRCh38
rs149686954 Chr.6: 63721563-63721563 on Build GRCh38
rs936322988 Chr.6: 63721467-63721467 on Build GRCh38
rs775519718 Chr.6: 63721549-63721549 on Build GRCh38
rs565204525 Chr.6: 63721543-63721543 on Build GRCh38
rs758638457 Chr.6: 63721454-63721454 on Build GRCh38
rs758984036 Chr.6: 63721474-63721474 on Build GRCh38
rs536900025 Chr.6: 63721586-63721586 on Build GRCh38
rs948892214 Chr.6: 63721420-63721420 on Build GRCh38
rs766969858 Chr.6: 63721475-63721475 on Build GRCh38
rs777582301 Chr.6: 63721585-63721585 on Build GRCh38
rs144513453 Chr.6: 63721602-63721602 on Build GRCh38
rs933169926 Chr.6: 63721425-63721425 on Build GRCh38
rs1009313420 Chr.6: 63721531-63721531 on Build GRCh38
rs751610535 Chr.6: 63721521-63721521 on Build GRCh38
rs892093060 Chr.6: 63721555-63721555 on Build GRCh38
rs1044679782 Chr.6: 63721444-63721444 on Build GRCh38
rs1024731403 Chr.6: 63721554-63721554 on Build GRCh38
rs1050742628 Chr.6: 63721433-63721433 on Build GRCh38
rs532257188 Chr.6: 63721544-63721544 on Build GRCh38
rs766405479 Chr.6: 63721448-63721448 on Build GRCh38

View Full Product Details