Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 59508644-59508917 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM81A
Entrez Gene ID
145773
Gene Name
family with sequence similarity 81 member A
Gene Aliases
-
UniGene
Hs.531168
Gene Chromosome Location
Chr.15: 59399662-59523555 on Build GRCh38
Transcripts
NM_152450.2 XM_005254166.2 XM_006720398.3 XM_006720399.2 XM_011521247.2 XM_011521248.2 XM_011521249.2 XM_011521250.2 XM_017021931.1 XM_017021932.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2219804 c.569A>T p.D190V Chr.15: 59508888-59508888 on Build GRCh38
5983971 c.581C>G p.S194* Chr.15: 59508900-59508900 on Build GRCh38
6015803 c.550C>T p.Q184* Chr.15: 59508869-59508869 on Build GRCh38
7367509 c.568G>T p.D190Y Chr.15: 59508887-59508887 on Build GRCh38
4340461 c.578T>C p.I193T Chr.15: 59508897-59508897 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748283158 Chr.15: 59508831-59508831 on Build GRCh38
rs371602814 Chr.15: 59508825-59508825 on Build GRCh38
rs367779634 Chr.15: 59508813-59508813 on Build GRCh38
rs972942026 Chr.15: 59508747-59508747 on Build GRCh38
rs777813611 Chr.15: 59508839-59508839 on Build GRCh38
rs779519422 Chr.15: 59508687-59508687 on Build GRCh38
rs561253138 Chr.15: 59508688-59508688 on Build GRCh38
rs758073691 Chr.15: 59508826-59508826 on Build GRCh38
rs371866554 Chr.15: 59508798-59508799 on Build GRCh38
rs774780213 Chr.15: 59508854-59508854 on Build GRCh38
rs894807049 Chr.15: 59508678-59508678 on Build GRCh38
rs759965552 Chr.15: 59508859-59508859 on Build GRCh38
rs970560985 Chr.15: 59508733-59508733 on Build GRCh38
rs763666855 Chr.15: 59508814-59508814 on Build GRCh38
rs753875873 Chr.15: 59508818-59508818 on Build GRCh38
rs772283561 Chr.15: 59508860-59508860 on Build GRCh38
rs569754527 Chr.15: 59508673-59508673 on Build GRCh38
rs778919977 Chr.15: 59508824-59508824 on Build GRCh38
rs761342077 Chr.15: 59508889-59508889 on Build GRCh38
rs182707407 Chr.15: 59508851-59508851 on Build GRCh38
rs189929963 Chr.15: 59508835-59508835 on Build GRCh38
rs546827419 Chr.15: 59508849-59508849 on Build GRCh38
rs541262727 Chr.15: 59508669-59508670 on Build GRCh38
rs781502131 Chr.15: 59508828-59508828 on Build GRCh38
rs755807688 Chr.15: 59508675-59508675 on Build GRCh38
rs757249186 Chr.15: 59508822-59508822 on Build GRCh38
rs775248689 Chr.15: 59508865-59508865 on Build GRCh38
rs748816256 Chr.15: 59508731-59508731 on Build GRCh38
rs760222128 Chr.15: 59508880-59508880 on Build GRCh38
rs763755156 Chr.15: 59508884-59508884 on Build GRCh38

View Full Product Details