Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66564445-66564718 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSF
Entrez Gene ID
8722
Gene Name
cathepsin F
Gene Aliases
CATSF, CLN13
UniGene
Hs.11590
Gene Chromosome Location
Chr.11: 66563463-66568576 on Build GRCh38
Transcripts
NM_003793.3 XM_011545328.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
169726 c.1252C>T p.R418C Chr.11: 66564627-66564627 on Build GRCh38
3452117 c.1248C>T p.I416I Chr.11: 66564631-66564631 on Build GRCh38
2040450 c.1231T>C p.F411L Chr.11: 66564648-66564648 on Build GRCh38
7198282 c.1320C>T p.N440N Chr.11: 66564559-66564559 on Build GRCh38
6194344 c.1242C>T p.H414H Chr.11: 66564637-66564637 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1005776027 Chr.11: 66564517-66564517 on Build GRCh38
rs755154475 Chr.11: 66564566-66564566 on Build GRCh38
rs200426008 Chr.11: 66564636-66564636 on Build GRCh38
rs150922871 Chr.11: 66564558-66564558 on Build GRCh38
rs752911567 Chr.11: 66564655-66564655 on Build GRCh38
rs768570314 Chr.11: 66564676-66564676 on Build GRCh38
rs199830949 Chr.11: 66564661-66564661 on Build GRCh38
rs201295932 Chr.11: 66564617-66564617 on Build GRCh38
rs779164010 Chr.11: 66564642-66564642 on Build GRCh38
rs777763116 Chr.11: 66564659-66564661 on Build GRCh38
rs778969360 Chr.11: 66564583-66564583 on Build GRCh38
rs368422217 Chr.11: 66564549-66564549 on Build GRCh38
rs571987848 Chr.11: 66564502-66564502 on Build GRCh38
rs867917295 Chr.11: 66564570-66564570 on Build GRCh38
rs747078300 Chr.11: 66564668-66564668 on Build GRCh38
rs759093942 Chr.11: 66564637-66564637 on Build GRCh38
rs781257663 Chr.11: 66564600-66564600 on Build GRCh38
rs770919773 Chr.11: 66564682-66564684 on Build GRCh38
rs761039140 Chr.11: 66564627-66564627 on Build GRCh38
rs1021882738 Chr.11: 66564576-66564577 on Build GRCh38
rs534376817 Chr.11: 66564557-66564557 on Build GRCh38
rs985016814 Chr.11: 66564532-66564532 on Build GRCh38
rs772558572 Chr.11: 66564693-66564693 on Build GRCh38
rs141345438 Chr.11: 66564626-66564626 on Build GRCh38
rs370667878 Chr.11: 66564663-66564663 on Build GRCh38
rs758542993 Chr.11: 66564656-66564656 on Build GRCh38
rs745759723 Chr.11: 66564603-66564603 on Build GRCh38
rs748168574 Chr.11: 66564584-66564584 on Build GRCh38
rs933983229 Chr.11: 66564596-66564596 on Build GRCh38
rs373027690 Chr.11: 66564641-66564642 on Build GRCh38
rs769678799 Chr.11: 66564610-66564610 on Build GRCh38
rs971072056 Chr.11: 66564469-66564469 on Build GRCh38
rs772030326 Chr.11: 66564593-66564593 on Build GRCh38
rs775373144 Chr.11: 66564613-66564613 on Build GRCh38
rs28464796 Chr.11: 66564618-66564618 on Build GRCh38
rs145087378 Chr.11: 66564641-66564641 on Build GRCh38
rs113541570 Chr.11: 66564675-66564675 on Build GRCh38
rs367661681 Chr.11: 66564692-66564692 on Build GRCh38
rs768131664 Chr.11: 66564540-66564540 on Build GRCh38
rs761351900 Chr.11: 66564685-66564685 on Build GRCh38
rs1001199791 Chr.11: 66564511-66564511 on Build GRCh38
rs140795906 Chr.11: 66564564-66564564 on Build GRCh38
rs762721365 Chr.11: 66564614-66564614 on Build GRCh38
rs980109275 Chr.11: 66564662-66564662 on Build GRCh38
rs375050229 Chr.11: 66564522-66564522 on Build GRCh38
rs1034531584 Chr.11: 66564513-66564513 on Build GRCh38
rs752410509 Chr.11: 66564664-66564664 on Build GRCh38
rs149687246 Chr.11: 66564565-66564565 on Build GRCh38
rs1055013673 Chr.11: 66564634-66564634 on Build GRCh38
rs768353764 Chr.11: 66564616-66564616 on Build GRCh38
rs774192403 Chr.11: 66564684-66564684 on Build GRCh38
rs779996406 Chr.11: 66564672-66564672 on Build GRCh38
rs1044709939 Chr.11: 66564496-66564496 on Build GRCh38
rs553298368 Chr.11: 66564629-66564629 on Build GRCh38

View Full Product Details