Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 109972176-109972449 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNV1
Entrez Gene ID
27012
Gene Name
potassium voltage-gated channel modifier subfamily V member 1
Gene Aliases
HNKA, KCNB3, KV2.3, KV8.1
UniGene
Hs.13285
Gene Chromosome Location
Chr.8: 109963645-109975847 on Build GRCh38
Transcripts
NM_014379.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
312229 c.805C>T p.R269C Chr.8: 109972444-109972444 on Build GRCh38
3644131 c.800G>A p.R267K Chr.8: 109972449-109972449 on Build GRCh38
6860810 c.948G>T p.L316L Chr.8: 109972301-109972301 on Build GRCh38
1454111 c.991G>T p.G331* Chr.8: 109972258-109972258 on Build GRCh38
1454112 c.899C>T p.T300I Chr.8: 109972350-109972350 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761024075 Chr.8: 109972388-109972388 on Build GRCh38
rs760934313 Chr.8: 109972259-109972259 on Build GRCh38
rs200841067 Chr.8: 109972225-109972225 on Build GRCh38
rs753266575 Chr.8: 109972427-109972427 on Build GRCh38
rs747782041 Chr.8: 109972324-109972324 on Build GRCh38
rs879256001 Chr.8: 109972287-109972287 on Build GRCh38
rs553228200 Chr.8: 109972330-109972330 on Build GRCh38
rs780655637 Chr.8: 109972319-109972319 on Build GRCh38
rs537206465 Chr.8: 109972206-109972206 on Build GRCh38
rs192812890 Chr.8: 109972288-109972288 on Build GRCh38
rs774275740 Chr.8: 109972349-109972349 on Build GRCh38
rs866022506 Chr.8: 109972358-109972358 on Build GRCh38
rs759980426 Chr.8: 109972424-109972424 on Build GRCh38
rs766834981 Chr.8: 109972396-109972396 on Build GRCh38
rs754367305 Chr.8: 109972280-109972280 on Build GRCh38
rs765796716 Chr.8: 109972426-109972426 on Build GRCh38
rs573160350 Chr.8: 109972337-109972337 on Build GRCh38
rs756799710 Chr.8: 109972316-109972316 on Build GRCh38
rs144174724 Chr.8: 109972309-109972309 on Build GRCh38
rs188651942 Chr.8: 109972271-109972271 on Build GRCh38
rs779587418 Chr.8: 109972293-109972293 on Build GRCh38
rs773302507 Chr.8: 109972385-109972385 on Build GRCh38
rs938796477 Chr.8: 109972291-109972291 on Build GRCh38
rs1014006283 Chr.8: 109972249-109972249 on Build GRCh38
rs1007659998 Chr.8: 109972386-109972386 on Build GRCh38
rs377022162 Chr.8: 109972242-109972242 on Build GRCh38
rs1017775576 Chr.8: 109972416-109972416 on Build GRCh38
rs149785439 Chr.8: 109972352-109972352 on Build GRCh38
rs772251494 Chr.8: 109972368-109972368 on Build GRCh38
rs746596 Chr.8: 109972243-109972243 on Build GRCh38
rs952312115 Chr.8: 109972382-109972382 on Build GRCh38
rs374913273 Chr.8: 109972421-109972421 on Build GRCh38
rs768538149 Chr.8: 109972346-109972346 on Build GRCh38
rs1027851141 Chr.8: 109972363-109972363 on Build GRCh38
rs754195590 Chr.8: 109972420-109972420 on Build GRCh38
rs139784477 Chr.8: 109972320-109972320 on Build GRCh38

View Full Product Details