Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 5404485-5404756 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FARS2
Entrez Gene ID
10667
Gene Name
phenylalanyl-tRNA synthetase 2, mitochondrial
Gene Aliases
COXPD14, FARS1, HSPC320, PheRS
UniGene
Hs.484547
Gene Chromosome Location
Chr.6: 5261001-5771592 on Build GRCh38
Transcripts
NM_001318872.1 NM_006567.4 XM_005248812.3 XM_006714966.2 XM_011514247.2 XM_011514248.2 XM_011514249.1 XM_011514251.2 XM_017010186.1 XM_017010187.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
365342 c.702G>T p.E234D Chr.6: 5404631-5404631 on Build GRCh38
7077591 c.707T>G p.V236G Chr.6: 5404636-5404636 on Build GRCh38
3697910 c.724G>C p.D242H Chr.6: 5404653-5404653 on Build GRCh38
3874835 c.706G>A p.V236M Chr.6: 5404635-5404635 on Build GRCh38
4683430 c.693C>T p.H231H Chr.6: 5404622-5404622 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752801499 Chr.6: 5404664-5404664 on Build GRCh38
rs951916099 Chr.6: 5404679-5404679 on Build GRCh38
rs377582332 Chr.6: 5404681-5404681 on Build GRCh38
rs767530718 Chr.6: 5404659-5404659 on Build GRCh38
rs751662656 Chr.6: 5404558-5404558 on Build GRCh38
rs555096870 Chr.6: 5404550-5404550 on Build GRCh38
rs201991648 Chr.6: 5404605-5404605 on Build GRCh38
rs114630373 Chr.6: 5404726-5404726 on Build GRCh38
rs754862565 Chr.6: 5404670-5404671 on Build GRCh38
rs771267489 Chr.6: 5404721-5404721 on Build GRCh38
rs746480149 Chr.6: 5404702-5404702 on Build GRCh38
rs770287901 Chr.6: 5404704-5404704 on Build GRCh38
rs773648691 Chr.6: 5404710-5404710 on Build GRCh38
rs190712413 Chr.6: 5404625-5404625 on Build GRCh38
rs756084801 Chr.6: 5404578-5404578 on Build GRCh38
rs748159371 Chr.6: 5404572-5404572 on Build GRCh38
rs375439106 Chr.6: 5404533-5404533 on Build GRCh38
rs763500245 Chr.6: 5404715-5404715 on Build GRCh38
rs758640166 Chr.6: 5404548-5404548 on Build GRCh38
rs895933196 Chr.6: 5404521-5404521 on Build GRCh38
rs7759327 Chr.6: 5404725-5404725 on Build GRCh38
rs370034437 Chr.6: 5404604-5404604 on Build GRCh38
rs371390356 Chr.6: 5404683-5404683 on Build GRCh38
rs1044543244 Chr.6: 5404567-5404567 on Build GRCh38
rs766636921 Chr.6: 5404634-5404634 on Build GRCh38
rs201092264 Chr.6: 5404623-5404623 on Build GRCh38
rs202060864 Chr.6: 5404596-5404596 on Build GRCh38
rs369015058 Chr.6: 5404635-5404635 on Build GRCh38
rs561270136 Chr.6: 5404516-5404516 on Build GRCh38
rs754954575 Chr.6: 5404565-5404565 on Build GRCh38
rs745577747 Chr.6: 5404687-5404687 on Build GRCh38
rs745400923 Chr.6: 5404603-5404603 on Build GRCh38
rs777757854 Chr.6: 5404589-5404589 on Build GRCh38
rs1009933151 Chr.6: 5404630-5404630 on Build GRCh38
rs766470412 Chr.6: 5404552-5404552 on Build GRCh38
rs958280596 Chr.6: 5404598-5404598 on Build GRCh38
rs759795755 Chr.6: 5404644-5404644 on Build GRCh38
rs773811639 Chr.6: 5404601-5404601 on Build GRCh38
rs781314140 Chr.6: 5404571-5404571 on Build GRCh38
rs763234952 Chr.6: 5404633-5404633 on Build GRCh38
rs770650914 Chr.6: 5404597-5404597 on Build GRCh38
rs777815118 Chr.6: 5404667-5404667 on Build GRCh38
rs1047260982 Chr.6: 5404684-5404684 on Build GRCh38
rs779590953 Chr.6: 5404700-5404700 on Build GRCh38
rs764360302 Chr.6: 5404554-5404554 on Build GRCh38
rs116567033 Chr.6: 5404666-5404666 on Build GRCh38
rs200003967 Chr.6: 5404697-5404697 on Build GRCh38
rs137910005 Chr.6: 5404621-5404621 on Build GRCh38
rs753691420 Chr.6: 5404680-5404680 on Build GRCh38
rs762348813 Chr.6: 5404611-5404611 on Build GRCh38

View Full Product Details