Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 48499887-48500160 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRYL
Entrez Gene ID
285527
Gene Name
FRY like transcription coactivator
Gene Aliases
AF4p12, KIAA0826, MOR2
UniGene
Hs.595553
Gene Chromosome Location
Chr.4: 48497363-48780312 on Build GRCh38
Transcripts
NM_015030.1 XM_005248090.4 XM_005248093.4 XM_011513680.2 XM_011513681.2 XM_011513682.2 XM_011513683.2 XM_011513684.2 XM_017008046.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2849073 c.8783G>C p.R2928T Chr.4: 48500030-48500030 on Build GRCh38
7191966 c.8657C>A p.S2886Y Chr.4: 48500156-48500156 on Build GRCh38
5014422 c.8764G>A p.A2922T Chr.4: 48500049-48500049 on Build GRCh38
261027 c.8746G>T p.E2916* Chr.4: 48500067-48500067 on Build GRCh38
5673213 c.8763A>T p.V2921V Chr.4: 48500050-48500050 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs181257019 Chr.4: 48499979-48499979 on Build GRCh38
rs571789225 Chr.4: 48499934-48499934 on Build GRCh38
rs188385001 Chr.4: 48499970-48499970 on Build GRCh38
rs781275636 Chr.4: 48500133-48500133 on Build GRCh38
rs769494934 Chr.4: 48500006-48500006 on Build GRCh38
rs918355082 Chr.4: 48500111-48500111 on Build GRCh38
rs747572215 Chr.4: 48499985-48499988 on Build GRCh38
rs745907238 Chr.4: 48500135-48500135 on Build GRCh38
rs540689433 Chr.4: 48500087-48500087 on Build GRCh38
rs747222952 Chr.4: 48500079-48500079 on Build GRCh38
rs753277835 Chr.4: 48500053-48500053 on Build GRCh38
rs546254706 Chr.4: 48500018-48500018 on Build GRCh38
rs758794120 Chr.4: 48500059-48500059 on Build GRCh38
rs954099936 Chr.4: 48499976-48499976 on Build GRCh38
rs765789722 Chr.4: 48500004-48500004 on Build GRCh38
rs760365675 Chr.4: 48500001-48500001 on Build GRCh38
rs779417959 Chr.4: 48500023-48500023 on Build GRCh38
rs185751021 Chr.4: 48500071-48500071 on Build GRCh38
rs912418218 Chr.4: 48499987-48499987 on Build GRCh38
rs771251980 Chr.4: 48500112-48500112 on Build GRCh38
rs569528521 Chr.4: 48500016-48500016 on Build GRCh38

View Full Product Details