Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 45464942-45465203 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COG3
Entrez Gene ID
83548
Gene Name
component of oligomeric golgi complex 3
Gene Aliases
SEC34
UniGene
Hs.507948
Gene Chromosome Location
Chr.13: 45464895-45536698 on Build GRCh38
Transcripts
NM_031431.3 XM_011535266.1 XM_011535267.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
947647 c.153G>T p.E51D Chr.13: 45465189-45465189 on Build GRCh38
5842571 c.166C>T p.P56S Chr.13: 45465202-45465202 on Build GRCh38
5842570 c.148G>A p.A50T Chr.13: 45465184-45465184 on Build GRCh38
5648466 c.113G>A p.R38K Chr.13: 45465149-45465149 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs538009116 Chr.13: 45465068-45465069 on Build GRCh38
rs542249909 Chr.13: 45464968-45464968 on Build GRCh38
rs757274309 Chr.13: 45465116-45465116 on Build GRCh38
rs755933277 Chr.13: 45465145-45465145 on Build GRCh38
rs34678243 Chr.13: 45465018-45465018 on Build GRCh38
rs576050376 Chr.13: 45464997-45464997 on Build GRCh38
rs200180168 Chr.13: 45465071-45465071 on Build GRCh38
rs748184991 Chr.13: 45465027-45465027 on Build GRCh38
rs772040389 Chr.13: 45465162-45465162 on Build GRCh38
rs759235670 Chr.13: 45465077-45465077 on Build GRCh38
rs774381979 Chr.13: 45465002-45465002 on Build GRCh38
rs564733357 Chr.13: 45465082-45465082 on Build GRCh38
rs773525363 Chr.13: 45465036-45465036 on Build GRCh38
rs974350205 Chr.13: 45464980-45464980 on Build GRCh38
rs941797147 Chr.13: 45465022-45465022 on Build GRCh38
rs763660050 Chr.13: 45465114-45465114 on Build GRCh38
rs762490827 Chr.13: 45465099-45465099 on Build GRCh38
rs373457132 Chr.13: 45465131-45465131 on Build GRCh38
rs770894032 Chr.13: 45464989-45464989 on Build GRCh38
rs141222488 Chr.13: 45465155-45465155 on Build GRCh38
rs900671515 Chr.13: 45465136-45465136 on Build GRCh38
rs766640714 Chr.13: 45465047-45465047 on Build GRCh38
rs557353471 Chr.13: 45464988-45464988 on Build GRCh38
rs760744868 Chr.13: 45465041-45465041 on Build GRCh38
rs752423674 Chr.13: 45465097-45465097 on Build GRCh38
rs902813181 Chr.13: 45465048-45465048 on Build GRCh38
rs919760306 Chr.13: 45464984-45464984 on Build GRCh38
rs749511290 Chr.13: 45464992-45464992 on Build GRCh38
rs961264961 Chr.13: 45464969-45464969 on Build GRCh38
rs377320049 Chr.13: 45465030-45465030 on Build GRCh38
rs1042019094 Chr.13: 45465042-45465042 on Build GRCh38
rs958342295 Chr.13: 45465106-45465106 on Build GRCh38
rs111818704 Chr.13: 45465091-45465091 on Build GRCh38
rs751444143 Chr.13: 45465115-45465115 on Build GRCh38
rs764947212 Chr.13: 45465085-45465085 on Build GRCh38
rs1030432576 Chr.13: 45465161-45465161 on Build GRCh38
rs917639695 Chr.13: 45465006-45465006 on Build GRCh38
rs777968918 Chr.13: 45465169-45465169 on Build GRCh38
rs781104021 Chr.13: 45464996-45464996 on Build GRCh38
rs528692223 Chr.13: 45465143-45465143 on Build GRCh38
rs1054413377 Chr.13: 45465119-45465119 on Build GRCh38
rs762230625 Chr.13: 45465043-45465066 on Build GRCh38
rs543312923 Chr.13: 45465178-45465178 on Build GRCh38
rs997687835 Chr.13: 45465065-45465065 on Build GRCh38
rs768914260 Chr.13: 45465001-45465001 on Build GRCh38

View Full Product Details