Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 33593434-33593699 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP26
Entrez Gene ID
78986
Gene Name
dual specificity phosphatase 26 (putative)
Gene Aliases
DSP-4, DUSP24, LDP-4, MKP-8, MKP8, NATA1, NEAP, SKRP3
UniGene
Hs.8719
Gene Chromosome Location
Chr.8: 33591330-33600106 on Build GRCh38
Transcripts
NM_001305115.1 NM_001305116.1 NM_024025.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3648583 c.370T>A p.F124I Chr.8: 33593599-33593599 on Build GRCh38
2959234 c.335G>A p.R112H Chr.8: 33593634-33593634 on Build GRCh38
360422 c.403G>T p.D135Y Chr.8: 33593566-33593566 on Build GRCh38
4752422 c.281C>T p.A94V Chr.8: 33593688-33593688 on Build GRCh38
3834612 c.414C>T p.H138H Chr.8: 33593555-33593555 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147101666 Chr.8: 33593566-33593566 on Build GRCh38
rs766538306 Chr.8: 33593549-33593549 on Build GRCh38
rs376826832 Chr.8: 33593554-33593554 on Build GRCh38
rs745572276 Chr.8: 33593486-33593486 on Build GRCh38
rs936137161 Chr.8: 33593623-33593623 on Build GRCh38
rs776386890 Chr.8: 33593634-33593634 on Build GRCh38
rs369900463 Chr.8: 33593593-33593593 on Build GRCh38
rs772301556 Chr.8: 33593677-33593677 on Build GRCh38
rs781725072 Chr.8: 33593661-33593661 on Build GRCh38
rs570972362 Chr.8: 33593502-33593502 on Build GRCh38
rs2044162 Chr.8: 33593508-33593508 on Build GRCh38
rs754649348 Chr.8: 33593660-33593660 on Build GRCh38
rs778281252 Chr.8: 33593484-33593484 on Build GRCh38
rs141858173 Chr.8: 33593670-33593670 on Build GRCh38
rs753031331 Chr.8: 33593662-33593662 on Build GRCh38
rs11545083 Chr.8: 33593639-33593639 on Build GRCh38
rs138179742 Chr.8: 33593663-33593663 on Build GRCh38
rs995253444 Chr.8: 33593499-33593499 on Build GRCh38
rs762384016 Chr.8: 33593655-33593655 on Build GRCh38
rs141867389 Chr.8: 33593553-33593553 on Build GRCh38
rs749329952 Chr.8: 33593611-33593611 on Build GRCh38
rs112579319 Chr.8: 33593472-33593472 on Build GRCh38
rs763915925 Chr.8: 33593633-33593633 on Build GRCh38
rs1009333379 Chr.8: 33593652-33593652 on Build GRCh38
rs898264585 Chr.8: 33593478-33593478 on Build GRCh38
rs746603542 Chr.8: 33593574-33593574 on Build GRCh38
rs11989709 Chr.8: 33593612-33593612 on Build GRCh38
rs7008370 Chr.8: 33593567-33593567 on Build GRCh38
rs750309974 Chr.8: 33593642-33593642 on Build GRCh38
rs749924912 Chr.8: 33593568-33593568 on Build GRCh38
rs769986511 Chr.8: 33593512-33593512 on Build GRCh38
rs778152331 Chr.8: 33593664-33593664 on Build GRCh38
rs150240540 Chr.8: 33593601-33593601 on Build GRCh38
rs770182203 Chr.8: 33593602-33593602 on Build GRCh38
rs780595244 Chr.8: 33593503-33593504 on Build GRCh38
rs201566430 Chr.8: 33593656-33593656 on Build GRCh38
rs138833176 Chr.8: 33593606-33593606 on Build GRCh38
rs35266053 Chr.8: 33593480-33593481 on Build GRCh38
rs757963284 Chr.8: 33593498-33593498 on Build GRCh38
rs775743791 Chr.8: 33593679-33593679 on Build GRCh38
rs143679528 Chr.8: 33593629-33593629 on Build GRCh38
rs771064956 Chr.8: 33593671-33593671 on Build GRCh38
rs763037611 Chr.8: 33593526-33593526 on Build GRCh38
rs147770891 Chr.8: 33593550-33593550 on Build GRCh38
rs929424539 Chr.8: 33593561-33593561 on Build GRCh38
rs779806348 Chr.8: 33593501-33593501 on Build GRCh38
rs757886243 Chr.8: 33593569-33593569 on Build GRCh38
rs905144370 Chr.8: 33593613-33593613 on Build GRCh38
rs773136621 Chr.8: 33593525-33593525 on Build GRCh38
rs755455913 Chr.8: 33593589-33593589 on Build GRCh38
rs765119983 Chr.8: 33593640-33593640 on Build GRCh38
rs778932566 Chr.8: 33593676-33593676 on Build GRCh38
rs751327813 Chr.8: 33593659-33593659 on Build GRCh38
rs774610495 Chr.8: 33593624-33593624 on Build GRCh38
rs761618847 Chr.8: 33593635-33593635 on Build GRCh38
rs114385466 Chr.8: 33593573-33593573 on Build GRCh38
rs753839287 Chr.8: 33593558-33593558 on Build GRCh38

View Full Product Details